MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Astrocyte activation and neurotoxicity: A study in different rat brain regions and in rat C6 astroglial cells.
Astrocyte activation and neurotoxicity: A study in different rat brain regions and in rat C6 astroglial cells.
Environmental toxicology and pharmacology (2015-06-27)
Poonam Goswami, Sonam Gupta, Neeraj Joshi, Sharad Sharma, Sarika Singh
ABSTRACT
The present study was conducted to investigate the effect of rotenone on astrocytes activation, their viability and its effect on neuronal death in different brain regions. Rotenone was injected in rat brain by intracerebroventricularly (bilateral) route at dose of 6 μg and 12 μg. In vitro C6 cells were treated with rotenone at concentration of 0.1, 0.25, 0.5, 1 and 2 μM. Rotenone administration to rat brain caused significant astrocytes activation in frontal cortex, cerebellum, cerebellar nucleus, substantia nigra, hypothalamus and hippocampus regions of the rat brain. Rotenone administration also led to significant degeneration of cells in all the studied regions along with altered nuclear morphology assessed by hematoxylin-eosin and cresyl violet staining. Histological staining showed the significantly decreased number of cells in all the studied regions except cerebellar nucleus in dose and time dependant manner. Rotenone administration in the rat brain also caused significant decrease in glutathione levels and augmented nitrite levels. In vitro treatment of rotenone to astrocytic C6 cells caused significantly increased expression of glial fibrillar acidic protein (GFAP) and decreased viability in dose and time dependent manner. Rotenone treatment to C6 cells exhibited significant generation of reactive oxygen species, augmented nitrite level, impaired mitochondrial activity, apoptotic chromatin condensation and DNA damage in comparison to control cells. Findings showed that oxidative stress play a considerable role in rotenone induced astrocyte death that was attenuated with co-treatment of antioxidant melatonin. In conclusion, results showed that rotenone caused significant astrocytes activation, altered nuclear morphology, biochemical alteration and apoptotic cell death in different rat brain regions. In vitro observations in C6 cells showed that rotenone treatment exhibited oxidative stress mediated apoptotic cell death, which was attenuated with co treatment of melatonin.
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N