MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Activation of angiotensin II type 1 receptors and contractile activity in human sigmoid colon in vitro.
Activation of angiotensin II type 1 receptors and contractile activity in human sigmoid colon in vitro.
Acta physiologica (Oxford, England) (2015-06-09)
M Mastropaolo, M G Zizzo, M Auteri, G Caldara, R Liotta, F Mulè, R Serio
ABSTRACT
To analyse the effects of angiotensin II (Ang II) on the contractility of human sigmoid colon, and to characterize the subtype(s) of receptor(s) involved and the related action mechanism. The contractility of sigmoid colon circular muscle strips was recorded isometrically. RT-PCR and immunohistochemistry were used to reveal the eventual existence of a local renin-angiotensin system (RAS) and the distribution of Ang II receptors. Transcripts encoding for the Ang II type 1 (AT1 ) and the Ang II type 2 (AT2 ) receptor subtypes and for the angiotensin-converting enzyme in the whole-thickness muscular wall were observed. Ang II caused a concentration-dependent contractile response, which is antagonized by losartan, AT1 receptor antagonist, but not by PD123319, AT2 receptor antagonist. The joint application of losartan and PD123319 did not produce any additive effect. The contractile response to Ang II was partially reduced by tetrodotoxin, Na(+) voltage-gated neural channel blocker, and to some extent by SR48968, tachykinin NK2 receptor antagonist. However, hexamethonium, nicotinic receptor antagonist, atropine, cholinergic muscarinic receptor antagonist and SR140333, tachykinin NK1 receptor antagonist, were ineffective. Immunohistochemical analysis showed that AT1 receptors were expressed on the smooth muscle layers and myenteric plexus. Ang II positively modulates the spontaneous contractile activity of human sigmoid colon via activation of post-junctional and pre-junctional AT1 receptors, the latter located on the enteric nerves that modulate the release of tachykinins. The presence of the components of RAS in the human colon suggests that Ang II can be also locally generated to control colonic motility.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Benzamide, purified by sublimation, ≥99.5%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Hexamethonium bromide
Sigma-Aldrich
Benzamide, 99%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology