MilliporeSigma
  • Impact of Storage Conditions on the Stability of Volatile Sulfur Compounds in Sampling Bags.
Impact of Storage Conditions on the Stability of Volatile Sulfur Compounds in Sampling Bags.
Journal of environmental quality (2015-10-06)
Hung V Le, Eric C Sivret, Gavin Parcsi, Richard M Stuetz
ABSTRACT
Odorous emissions from agricultural and waste management operations can cause annoyance to local populations. Volatile sulfur compounds (VSCs) are dominant odorants that are often lost during collection using sample bags. The degree of VSC losses depends on factors such as storage time, bag materials, temperature, sample relative humidity (RH), light exposure, and the presence of volatile organic compounds (VOCs). To assess the impact of those factors on the stability of 10 VSCs (hydrogen sulfide, methanethiol, ethanethiol, dimethyl sulfide, tert-butanethiol, ethyl methyl sulfide, 1-butanethiol, dimethyl disulfide, diethyl disulfide, and dimethyl trisulfide), laboratory-based experiments were conducted according to a factorial experimental design. Linear mixed-effects models were constructed for loss predictions. The estimated recovery of HS in Tedlar bag was 8 to 10% higher than in Mylar and Nalophan between 6 and 30 h. At ≤20°C and without being exposed to light, at least 75% relative recovery of the 10 VSCs in Tedlar bags can be achieved after 18 h, whereas, a maximum of 12 h of storage should not be exceeded to ensure a minimum of 74% relative recovery of the VSCs in Mylar and Nalophan bags.
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl disulfide, ≥98%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Nitrogen, ≥99.998%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl disulfide, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Pricing and availability is not currently available.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Pricing and availability is not currently available.
Sigma-Aldrich
Rhein, technical grade
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanethiol, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Pricing and availability is not currently available.
Sigma-Aldrich
Rhein
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanethiol, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.