Skip to Content
MilliporeSigma

EMU011871

MISSION® esiRNA

targeting mouse Itgb2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$242.00
50 μG
$435.00

$242.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGACAACTCCAACCAGTTTCAGACAGAGGTCGGCAAGCAACTGATTTCCGGAAACCTGGACGCCCCTGAGGGTGGGCTGGATGCCATAATGCAAGTTGCTGCATGTCCGGAGGAAATTGGCTGGCGCAATGTCACGAGGCTGCTGGTGTTTGCCACAGACGATGGCTTCCACTTTGCTGGTGATGGCAAACTGGGTGCCATCCTGACCCCCAATGATGGCCGCTGCCACCTGGAGGATAACATGTACAAGAGGAGCAATGAGTTCGACTACCCATCCGTGGGTCAGCTGGCACACAAACTTTCCGAGAGCAACATCCAGCCCATCTTTGCGGTGACAAAGAAGATGGTGAAAACGTATGAGAAACTCACGGAGATCATCCCCAAGTCAGCAGTGGGGGAACTGTCTGACGACTCCAGCAAC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU019391EMU017091EMU051281
Ensembl | mouse accession no.

ENSMUSG00000000290

Ensembl | mouse accession no.

ENSMUSG00000062352

Ensembl | mouse accession no.

ENSMUSG00000022817

Ensembl | mouse accession no.

ENSMUSG00000027087

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Gene Information

mouse ... ITGB2(16414), Itgb2(16414)

Gene Information

mouse ... ITGB1BP1(16413), Itgb1bp1(16413)

Gene Information

mouse ... ITGB5(16419), Itgb5(16419)

Gene Information

mouse ... Itgav(16410)

esiRNA cDNA target sequence

CCGACAACTCCAACCAGTTTCAGACAGAGGTCGGCAAGCAACTGATTTCCGGAAACCTGGACGCCCCTGAGGGTGGGCTGGATGCCATAATGCAAGTTGCTGCATGTCCGGAGGAAATTGGCTGGCGCAATGTCACGAGGCTGCTGGTGTTTGCCACAGACGATGGCTTCCACTTTGCTGGTGATGGCAAACTGGGTGCCATCCTGACCCCCAATGATGGCCGCTGCCACCTGGAGGATAACATGTACAAGAGGAGCAATGAGTTCGACTACCCATCCGTGGGTCAGCTGGCACACAAACTTTCCGAGAGCAACATCCAGCCCATCTTTGCGGTGACAAAGAAGATGGTGAAAACGTATGAGAAACTCACGGAGATCATCCCCAAGTCAGCAGTGGGGGAACTGTCTGACGACTCCAGCAAC

esiRNA cDNA target sequence

TTCAAGATCCAGCACTGTCGCCAGCCTCGATACAGACTCCACTAAAAGCTCAGGACAGAGTAACAGCAATTTAGACACATGTGCTGAGTTCCGAATAAAGTATGTTGGTGCCATCGAGAAACTGGCCGTGTCTGAGGGGAAGAGCCTTGAAGGACCACTAGACCTGATAAATTACATAGATGTCGCCCAGCAAGATGGAAAGTTGCCTTTTGTGCCTTTGGAAGAGGAGTTCATTCTGGGCGTTTCTAAGTACGGCATAAAAGTCTCGACCACGGATCAGCATGGTGTTCTGCATAGGCATGCCCTGTATCTAATCATCCGGATGGTGTGTTACGATGACGGCCTGGGAGCTGGGAAAAGCTTGCTGGCACTCAAGACCACAGATGCAAGCAACGAAGAGTACAGCCTGTGGGTTTACCAGTGTAACAGCCTGGAGCA

esiRNA cDNA target sequence

GGTTTCGGGTCTTTTGTTGACAAGGACATCTCTCCTTTCTCCTACACGGCACCGAGATACCAGACCAATCCGTGTATTGGTTACAAGTTATTCCCCAACTGCGTCCCCTCCTTCGGGTTCCGGCATCTGCTGCCTCTCACAGACAGAGTCGACAGCTTCAACGAGGAAGTGAGGAAGCAGAGGGTGTCCCGGAACCGAGATGCCCCCGAGGGGGGGTTTGATGCGGTCCTCCAGGCTGCTGTCTGCAAGGAGAAGATCGGATGGCGAAAAGATGCTCTGCACTTGCTGGTGTTCACAACAGACGATGTGCCCCACATCGCACTGGATGGAAAACTGGGTGGCCTGGTCCAGCCCCACGATGGCCAGTGTCACCTGAATGAAGCCAATGAGTACACAGCCTCTAACCAGATGGACTATCCATCGCTTGCCTTG

esiRNA cDNA target sequence

GTCCGACCACCTCAAGAAGAGCAAGAACGAGAACAGCTTCAGCCTCATGAGAATGGCGAAGGAAACTCTGAAACCTGAGGTGCTTCTTAAGTGGCGCCACTCTGTCCGCTCCAAGTTCACCACCGTGCTGGTGATTTCAACATCTCTCCTGAAGAGTGAAATCCCAGGACTGTATTGCTGAGAGCACCCAGCAGTCTCAGTTTATCCACAAAATTGAAATGGATATTTGTCGACTTCTCTTTATGAATAAGTTCAGGTTACATTTACTTCGCCCACACTGACGTGTGTTCTTAGGGACTTAAGTGATGGAATTTCAAGTGATTACCTTTGTTCAGTGTACATAAGACAGACAGCACCGAGTCAGCACGTTACATAGCATAGTACCTCTTCAGTTACTGTTCCTAATTGACTGTGTGGGGTTTCTCTCTTCGGCGTTACGT

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service