MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A chemical proteomics approach for global analysis of lysine monomethylome profiling.
A chemical proteomics approach for global analysis of lysine monomethylome profiling.
Molecular & cellular proteomics : MCP (2014-12-17)
Zhixiang Wu, Zhongyi Cheng, Mingwei Sun, Xuelian Wan, Ping Liu, Tieming He, Minjia Tan, Yingming Zhao
ABSTRACT
Methylation of lysine residues on histone proteins is known to play an important role in chromatin structure and function. However, non-histone protein substrates of this modification remain largely unknown. An effective approach for system-wide analysis of protein lysine methylation, particularly lysine monomethylation, is lacking. Here we describe a chemical proteomics approach for global screening for monomethyllysine substrates, involving chemical propionylation of monomethylated lysine, affinity enrichment of the modified monomethylated peptides, and HPLC/MS/MS analysis. Using this approach, we identified with high confidence 446 lysine monomethylation sites in 398 proteins, including three previously unknown histone monomethylation marks, representing the largest data set of protein lysine monomethylation described to date. Our data not only confirms previously discovered lysine methylation substrates in the nucleus and spliceosome, but also reveals new substrates associated with diverse biological processes. This method hence offers a powerful approach for dynamic study of protein lysine monomethylation under diverse cellular conditions and in human diseases.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Urea, ≥99.0%
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Methionine, European Pharmacopoeia (EP) Reference Standard
DL-Methionine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%