MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now
HPLC Columns
A collection of various Supelco<sup>®</sup> HPLC and UHPLC columns scattered on a green surface. These columns, which are cylindrical with metallic finishes and equipped with fittings at both ends, represent a range of chromatography column technologies for different analytical needs. They vary in size and some have labeling that indicates specifications or model numbers.
Supelco® HPLC and UHPLC columns meet today’s demands of fast U/HPLC, LC-MS, and biopolymer separation, as well as regulated pharmacopeia and agency methods within various industries. Supelco® HPLC products are available with Fused-Core® particle technology, monolithic silica, and fully porous particles, including ultra-pure silica. In addition, porous graphitic carbon (PGC) particles, and polymeric particles are available for high pH stability.
Products
Showing 1-20 of 2010
Change View
Showing 1-20 of 2010
Compare
Product Number
Product Name
Product Description
Pricing
Porous Graphitic Carbon (PGC) Particles
Porous graphitic carbon (PGC) columns, like the Supel™ Carbon LC column, provide unique chromatographic properties that are unmatched by silica particle-packed columns.  The full pH range can be used as well as high temperature (> 200 °C), opening up new frontiers for chromatographers.
Superficially Porous Silica Particles (SPP)/Fused Core® Technology
Superficially porous silica columns deliver higher speed and efficiency than fully porous particles of the same size. Fused-Core® HPLC columns like Ascentis® Express and BIOshell™ can convert any HPLC system into a fast HPLC workhorse with maximum speed and performance.
Monolithic Silica
  • Monolithic, silica-based HPLC columns such as Chromolith® HPLC columns allow rapid separations at very low column backpressure with high matrix tolerance and extended column lifetime, thus making them ideal products for high throughput and cost-efficient analyses of matrix rich samples.
  • Chromolith® HPLC columns are made from monolithic silica with a bimodal pore structure having 2 µm macropores and 13 nm mesopores. This structure leads to various key advantages to the column.
  • Chromolith® WP columns are wide pore (300 Å) monolithic silica columns made up of a single continuous rod of high purity, porous silica. These columns have shown great potential compared to conventional columns with consistent performance over a longer lifetime. In addition to various RP phases, a Protein A affinity column and an epoxy column, that enable customer specific modifications, are  also available.
Fully Porous Silica Particles (FPP) Type B (High Purity Silica)
  • Purospher® STAR HPLC and UHPLC columns are best suited for the peak-tailing free separation of basic, neutral, and metal chelating samples. Their superior performance and stability allow maximum flexibility in method development with extended pH stability.
  • Discovery® HPLC columns are efficient for the analysis of biomolecules and small molecules. These columns have polar and non-polar bondings, with unique retention, selectivity, and no-bleed properties for LC-MS applications.
  • The Ascentis® bonded phases have a wide range of selectivity. Type B silica makes the column inert, stable, and reproducible. The high surface area of the silica is suitable for  preparative loading capacity.
  • SeQuant® HILIC HPLC columns (hydrophilic interaction liquid chromatography) carry densely bonded, zwitterionic functional groups. These HILIC columns are available in a variety of lengths, particle sizes, and pore dimensions while showing an exceptional reproducibility and robustness.
  • Sepax® HPLC columns are highly porous silica-based columns mainly utilized for peptides and protein analysis.
Fully Porous Silica Particles (FPP) Type A (Conventional Silica)
  • Supelcosil® silica-based HPLC column line has a wide range of bonded phase chemistries, with the particle sizes and column configurations ranging from microbore to preparative scale. These columns show great efficiency for polar, nonpolar, and charged analytes.
  • LiChrospher® and Superspher® HPLC columns are highly versatile and reliable, proven in thousands of established HPLC methods.
  • LiChrosorb® HPLC columns are packed with irregular silica particles ranging from analytical scale 5 µm to large particles sized for production scale.
  • TSKgel® HPLC and UHPLC columns are available for gel filtration, gel permeation, ion exchange, reversed-phase, hydrophobic interaction, hydrophilic interaction, and affinity chromatography. (only available in the US and Europe)
Fully Porous Polymeric Particles
  • apHera™ reversed-phase and amino HPLC columns are stable at the pH range 2-12 and are amenable to washing with alkaline solutions. They are ideal for the analysis of peptides, small proteins, carbohydrates, polar drugs, and organic acids.
  • Supelcogel® HPLC columns are packed with polystyrene-divinylbenzene co-polymer (PS-DVB) making them chemically stable. The two variations of PS-DVB co-polymer (6% and 8% cross-linked) maximize the separation process of the samples.
  • Hamilton Polymeric Reversed Phase HPLC columns are extremely inert, efficient, and durable, with high mechanical and thermal stability.
  • TSKgel® HPLC and UHPLC columns are available for the separation of proteins, peptides, and other biomolecules. The columns containing silica and methacrylate particles with different pore sizes can accommodate analytes with a wide range of molecular weights. (only available in the US and Europe)
Chiral HPLC Columns
We have a wide range of chiral HPLC columns for various applications.
Related Resources
Page 1 of 6
Tutorials
Learn how to connect HPLC columns and cartridges with the appropriate pre-columns, holders, and guard cartridges.
*Tosoh Bioscience columns are available in the following countries:
Albania
Andorra
Argentina
Armenia
Austria
Azerbaijan
Afghanistan
Algeria
Albania
Armenia
Azerbaijan
Belarus
Belgium
Belize
Bolivia
Bosnia & Herzegovina
Brazil
Bulgaria
Bahrain
Belarus
Bosnia-Herz
Botswana
Burkina-Faso
Canada
Chile
Colombia
Costa Rica
Croatia
Cuba Dominican Republic
Cyprus
Czechia
Cameron
Central Afr. Rep
Chad
Congo
Congo dem. Rep.
Croatia
Denmark
Djibouti
Ecuador
Egypt
El Salvador
Eritrea
Ethiopia
Estonia
Finland
France
French Guiana
Georgia
Germany
Greece
Gabon
Georgia
Ghana
Guatemala
Guyana
Haiti
Honduras
Hungary
Iceland
Ireland
Italy
Iraq
Israel
Kazakhstan
Kosovo
Kazakhstan
Kenya
Kosovo
Kuwait
Kyrgyzstan
Lebanon
Libya
Latvia
Liechtenstein
Lithuania
Luxembourg
Madagascar
Malawi
Malta
Mauretania
Mauritius
Mexico
Moldova
Monaco
Montenegro
Moldavia
Mongolia
Morocco
Mozambique
Nicaragua
Nigeria
Netherlands
North Macedonia
(formerly Macedonia)
Norway
Oman
Panama
Paraguay
Peru
Poland
Portugal
Puerto Rico
Qatar
Russia
Rwanda
Romania
Russia
San Marino
Serbia
Slovakia
Slovenia
Spain
Sweden
Switzerland
Saudi Arabia
Senegal
Serbia
Serbia-Montenegro
South Africa
Suriname
Tadzhikistan
Tanzania
Togo
Tunisia
Turkey
Ukraine
Uruguay
Turkmenistan
United Kingdom (UK)
United States of America
Uganda
Ukraine
Utd.Arab.Emir.
Uzbekistan
Vatican City
Venezuela
Yemen
Zambia
Zimbabwe