Merck
The immunoregulator soluble TACI is released by ADAM10 and reflects B cell activation in autoimmunity.
Journal of immunology (Baltimore, Md. : 1950) (2014-12-17)
Franziska S Hoffmann, Peer-Hendrik Kuhn, Sarah A Laurent, Stefanie M Hauck, Kerstin Berer, Simone A Wendlinger, Markus Krumbholz, Mohsen Khademi, Tomas Olsson, Martin Dreyling, Hans-Walter Pfister, Tobias Alexander, Falk Hiepe, Tania Kümpfel, Howard C Crawford, Hartmut Wekerle, Reinhard Hohlfeld, Stefan F Lichtenthaler, Edgar Meinl
ABSTRAKT
BAFF and a proliferation-inducing ligand (APRIL), which control B cell homeostasis, are therapeutic targets in autoimmune diseases. TACI-Fc (atacicept), a soluble fusion protein containing the extracellular domain of the BAFF-APRIL receptor TACI, was applied in clinical trials. However, disease activity in multiple sclerosis unexpectedly increased, whereas in systemic lupus erythematosus, atacicept was beneficial. In this study, we show that an endogenous soluble TACI (sTACI) exists in vivo. TACI proteolysis involved shedding by a disintegrin and metalloproteinase 10 releasing sTACI from activated B cells. The membrane-bound stub was subsequently cleaved by γ-secretase reducing ligand-independent signaling of the remaining C-terminal fragment. The shed ectodomain assembled ligand independently in a homotypic way. It functioned as a decoy receptor inhibiting BAFF- and APRIL-mediated B cell survival and NF-κB activation. We determined sTACI levels in autoimmune diseases with established hyperactivation of the BAFF-APRIL system. sTACI levels were elevated both in the cerebrospinal fluid of the brain-restricted autoimmune disease multiple sclerosis correlating with intrathecal IgG production, as well as in the serum of the systemic autoimmune disease systemic lupus erythematosus correlating with disease activity. Together, we show that TACI is sequentially processed by a disintegrin and metalloproteinase 10 and γ-secretase. The released sTACI is an immunoregulator that shares decoy functions with atacicept. It reflects systemic and compartmentalized B cell accumulation and activation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Tricine, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)