Merck
  • The small molecule C-6 is selectively cytotoxic against breast cancer cells and its biological action is characterized by mitochondrial defects and endoplasmic reticulum stress.
The small molecule C-6 is selectively cytotoxic against breast cancer cells and its biological action is characterized by mitochondrial defects and endoplasmic reticulum stress.
Breast cancer research : BCR (2014-11-27)
Rachel M Vaden, Keith M Gligorich, Ranjan Jana, Matthew S Sigman, Bryan E Welm
ABSTRAKT
The establishment of drug resistance following treatment with chemotherapeutics is strongly associated with poor clinical outcome in patients, and drugs that target chemoresistant tumors have the potential to increase patient survival. In an effort to identify biological pathways of chemoresistant breast cancers that can be targeted therapeutically, a small molecule screen utilizing metastatic patient-derived breast cancer cells was conducted; from this previous report, the cytotoxic small molecule, C-6, was identified for its ability to selectively kill aggressive breast cancer cells in a caspase-independent manner. Here, we describe the cellular and molecular pathways induced following C-6 treatment in both normal and breast cancer cell lines. Transcriptome analyses and protein expression experiments were used to measure endoplasmic reticulum (ER) stress following C-6 treatment. Studies utilizing transmission electron microscopy and metabolomic profiling were conducted to characterize mitochondrial morphology and function in C-6-treated cells. Oxygen consumption rates and oxidative stress were also measured in breast cancer and normal mammary epithelial cells following treatment with the small molecule. Finally, structural modifications were made to the molecule and potency and cancer selectivity were evaluated. Treatment with C-6 resulted in ER stress in both breast cancer cells and normal mammary epithelial cells. Gross morphological defects were observed in the mitochondria and these aberrations were associated with metabolic imbalances and a diminished capacity for respiration. Following treatment with C-6, oxidative stress was observed in three breast cancer cell lines but not in normal mammary epithelial cells. Finally, synthetic modifications made to the small molecule resulted in the identification of the structural components that contribute to C-6's cancer-selective phenotype. The data reported here implicate mitochondrial and ER stress as a component of C-6's biological activity and provide insight into non-apoptotic cell death mechanisms; targeting biological pathways that induce mitochondrial dysfunction and ER stress may offer new strategies for the development of therapeutics that are effective against chemoresistant breast cancers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Myxothiazol, from Myxococcus fulvus Mx f85, ≥98% (HPLC)
Sigma-Aldrich
Dioctan 2′,7′-dichlorodihydrofluoresceiny, ≥97%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)