Merck
  • Evaluation and comparison of a new DOTA and DTPA-bombesin agonist in vitro and in vivo in low and high GRPR expressing prostate and breast tumor models.
Evaluation and comparison of a new DOTA and DTPA-bombesin agonist in vitro and in vivo in low and high GRPR expressing prostate and breast tumor models.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2014-12-06)
Priscilla B Pujatti, Julie M Foster, Ciara Finucane, Chantelle D Hudson, Jerome C Burnet, Kerly F M Pasqualoto, Jair Mengatti, Stephen J Mather, Elaine B de Araújo, Jane K Sosabowski
ABSTRAKT
We evaluated and compared a new bombesin analog [Tyr-Gly5, Nle(14)]-BBN(6-14) conjugated to DOTA or DTPA and radiolabeled with In-111 in low and high GRPR expressing tumor models. Both peptides were radiolabeled with high radiochemical purity and specific activity. In vitro assays on T-47D, LNCaP and PC-3 cells showed that the affinity of peptides is similar and a higher binding and internalization of DOTA-peptide to PC-3 cells was observed. Both peptides could target PC-3 and LNCaP tumors in vivo and both tumor types could be visualized by microSPECT/CT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, suitable for suitability for complexometry, ≥99.0%