Merck
Hemin improves insulin sensitivity in skeletal muscle in high fat-fed mice.
Journal of pharmacological sciences (2014-10-25)
Tae-Jin Ju, Woo-Young Kwon, Yong-Woon Kim, Jong-Yeon Kim, Yong-Dae Kim, In-Kyu Lee, So-Young Park
ABSTRAKT
The present study examined whether hemin could prevent the development of high-fat diet-induced insulin resistance in the liver and skeletal muscle using a hyperinsulinemic-euglycemic clamp. A four-week high-fat feeding to mice increased the body weight, fat mass, and plasma levels of insulin and lipid, which were reduced by hemin. High-fat diet reduced whole body glucose uptake, which were increased by hemin. Insulin-stimulated hepatic glucose production (HGP) was increased by high-fat diet, but hemin had no significant effect on HGP. Skeletal muscle glucose uptake was reduced by high-fat diet, and hemin normalized the glucose uptake. High-fat diet increased triglyceride levels and mRNA levels of lipogenic enzymes, and decreased mRNA levels of enzymes involved in lipid β-oxidation, which was reversed by hemin. Phosphorylated AMP-activated protein kinase levels were increased in the skeletal muscle of high fat-fed hemin-injected mice. High-fat diet reduced mRNA levels of antioxidant enzymes and increased mRNA levels of inflammatory cytokines and nitrotyrosine levels, which was normalized by hemin in the skeletal muscle. However, hemin had no significant effect on these factors in the liver. These results suggest that hemin prevents the development of high-fat diet-induced insulin resistance by increased insulin sensitivity in the skeletal muscle.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dekstroza, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.