Merck
  • SKLB-M8 induces apoptosis through the AKT/mTOR signaling pathway in melanoma models and inhibits angiogenesis with decrease of ERK1/2 phosphorylation.
SKLB-M8 induces apoptosis through the AKT/mTOR signaling pathway in melanoma models and inhibits angiogenesis with decrease of ERK1/2 phosphorylation.
Journal of pharmacological sciences (2014-10-25)
Jingjing Wang, Zhuang Yang, Jiaolin Wen, Feng Ma, Fang Wang, Kun Yu, Minghai Tang, Wenshuang Wu, Yinfeng Dong, Xia Cheng, Chunlai Nie, Lijuan Chen
ABSTRAKT
SKLB-M8, a derivative of millepachine, showed significant anti-proliferative effects in melanoma cell lines. In this study, we investigated the anti-melanoma and anti-angiogenic activity of SKLB-M8 on three melanoma cell lines (A2058, CHL-1, and B16F10) and human umbilical vein endothelial cells (HUVECs). In vitro, SKLB-M8 showed anti-proliferative activity with IC50 values of 0.07, 0.25, and 0.88 μM in A2058, CHL-1, and B16F10 cell lines, respectively. Flow cytometory analysis showed that SKLB-M8 induced G2/M arrest in three melanoma cell lines, and western blotting demonstrated that SKLB-M8 down-regulated the expression of cdc2, up-regulated p53 in A2058 and CHL-1 cells, and triggered cell apoptosis through down-regulating AKT and phosphorylated mTOR (p-mTOR). SKLB-M8 also inhibited HUVEC proliferation, migration, invasion, and tube formation in vitro with the inhibition of phosphorylated ERK1/2 (p-ERK1/2). In vivo, alginate-encapsulated tumor cell assay revealed that SKLB-M8 suppressed B16F10 tumor angiogenesis. In CHL-1- and B16F10-tumor-bearing mouse models, SKLB-M8 inhibited tumor growth by oral treatment with less toxicity. CD31 immunofluoresence staining and caspase-3 immunohistochemistry indicated that SKLB-M8 inhibited melanoma tumor growth by targeting angiogenesis and inducing caspase3-dependent apoptosis. SKLB-M8 might be a potential anti-melanoma drug candidate.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%