Merck
Pretransplant replacement of donor liver grafts with recipient Kupffer cells attenuates liver graft rejection in rats.
Journal of gastroenterology and hepatology (2014-12-24)
Kosuke Endo, Tomohide Hori, Kanta Jobara, Toshiyuki Hata, Tatsuaki Tsuruyama, Shinji Uemoto
ABSTRAKT
Rejection of liver grafts is a difficult issue that has not been resolved. Preoperative replacement of liver cells in the graft with cells from the intended recipient may attenuate rejection. We investigated whether preoperative transplant of recipient bone marrow cells (BMCs) to the donor replaced liver allograft cells and attenuated rejection. We used a rat model of allogeneic liver transplant (LT) from Dark Agouti (DA) to Lewis (LEW) rats. In BMC group, DA rats received BMC transplants from LacZ-transgenic LEW rats at 1 week before LT. In the control group, DA rats received no preoperative treatment. We evaluated graft damage at 7 days after LT and the survival of the recipient rats. Rats in the BMC group experienced prolonged survival that was abrogated by the administration of gadolinium chloride to donors at 24 h before LT. Serum concentrations of total bilirubin and hyaluronic acid on day 7 were significantly lower in the BMC group, and histopathological analyses revealed that rejection of the liver graft was attenuated. X-gal staining and immunohistostaining of the liver graft revealed that BMCs engrafted in the sinusoidal space differentiated into Kupffer cells. Preoperative transplant of recipient BMCs to LT donors replaced donor Kupffer cells and attenuated post-LT rejection, indicating that this strategy may increase the success of LT.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
4-Bromomethyl-7-methoxycoumarin, 97%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
SAFC
Sodium deoxycholate