Merck
  • Cis-9, trans-11 and trans-10, cis-12 CLA mixture does not change body composition, induces insulin resistance and increases serum HDL cholesterol level in rats.
Cis-9, trans-11 and trans-10, cis-12 CLA mixture does not change body composition, induces insulin resistance and increases serum HDL cholesterol level in rats.
Journal of oleo science (2015-04-07)
Mariana Macedo de Almeida, Yamara Oliveira de Souza, Sheila Cristina Potente Dutra Luquetti, Céphora Maria Sabarense, José Otávio do Amaral Corrêa, Ellen Paula Santos da Conceição, Patrícia Cristina Lisboa, Egberto Gaspar de Moura, Sara Malaguti Andrade Soares, Ana Cristina Moura Gualberto, Jacy Gameiro, Marco Antônio Sundfeld da Gama, Fernando César Ferraz Lopes, Raúl Marcel González Garcia
ABSTRAKT
Synthetic supplements of conjugated linoleic acid (CLA) containing 50:50 mixture of cis-9, trans-11 and trans-10, cis-12 CLA isomers have been commercialized in some places for reducing body fat. However the safety of this CLA mixture is controversial and in some countries the CLA usage as food supplement is not authorized. Changes in insulinemic control and serum lipids profile are potential negative effects related to consumption of CLA mixture. The present study aimed to evaluate the effects of a diet containing mixture of cis-9, trans-11 and trans-10, cis-12 CLA on prevention of obesity risk as well as on potential side effects such as insulin resistance and dyslipidemia in Wistar rats. Thirty male Wistar rats were randomly assigned to the following dietary treatments (n=10/group), for 60 days: Normolipidic Control (NC), diet containing 4.0% soybean oil (SO); High Fat-Control (HF-C), diet containing 24.0% SO; High Fat-synthetic CLA (HF-CLA), diet containing 1.5% of an isomeric CLA mixture (Luta-CLA 60) and 22.5% SO. Luta-CLA 60 (BASF) contained nearly 60% of CLA (cis-9, trans-11 and trans-10, cis-12 CLA at 50:50 ratio). The HF-CLA diet contained 0.3% of each CLA isomer. HF-CLA diet had no effect on dietary intake and body composition. HF-CLA-fed rats had lower levels of PPARγ protein in retroperitoneal adipose tissue, hyperinsulinemia compared to HF-C-fed rats, hyperglycemia compared to NC-fed rats while no differences in glycemia were observed between NC and HF-C groups, increased HOMA index and higher levels of serum HDL cholesterol. Thus, feeding rats with a high fat diet containing equal parts of cis-9, trans-11 and trans-10, cis-12 CLA isomers had no effect on body composition and induced insulin resistance. Despite HF-CLA-fed rats had increased serum HDL cholesterol levels, caution should be taken before synthetic supplements containing cis-9, trans-11 and trans-10, cis-12 CLA are recommended as a nutritional strategy for weight management.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder