Merck
Towards a feasible and scalable production of bio-xerogels.
Journal of colloid and interface science (2015-06-30)
Natalia Rey-Raap, Andrzej Szczurek, Vanessa Fierro, J Angel Menéndez, Ana Arenillas, Alain Celzard
ABSTRAKT
The synthesis process of carbon xerogels is limited, mainly due to two drawbacks that prevent their introduction onto the market: (i) the long time required for producing the material and (ii) the reagents used for the synthesis, which are costly and harmful to the environment. Microwave radiation is expected to produce a reduction in time of more than 90%, while the use of tannin instead of resorcinol will probably result in a cost-effective carbonaceous material. Resorcinol-tannin-formaldehyde xerogels containing different amounts of tannin, either with or without a surfactant (sodium dodecyl sulphate), were synthesized by means of two different heating methods: conventional and microwave heating. The effects of the surfactant, the heating method and the addition of tannin upon the porous structure and the chemical composition of the final materials were evaluated. It was found that the addition of surfactant is essential for obtaining highly porous xerogels when using tannins. The heating method also plays an important role, as conventionally synthesized samples display a greater volume of large pores. However, tannins are less sensitive to microwave radiation and their use results in tannin-formaldehyde xerogels that have a porous structure and chemical composition similar to those of resorcinol-formaldehyde xerogels.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Resorcinol, meets analytical specification of Ph. Eur., BP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Resorcinol, BioXtra, ≥99%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C