Merck
Optimization of the IL-8 Luc assay as an in vitro test for skin sensitization.
Toxicology in vitro : an international journal published in association with BIBRA (2015-07-19)
Yutaka Kimura, Chizu Fujimura, Yumiko Ito, Toshiya Takahashi, Yoshihiro Nakajima, Yoshihiro Ohmiya, Setsuya Aiba
ABSTRAKT
We previously reported a dataset of the IL-8 Luc assay covering reference chemicals published by ECVAM, in which the effects of chemicals on IL-8 promoter activity were evaluated by an IL-8 reporter cell line, THP-G8 cells. To clarify its performance, we created another dataset of 88 sensitizers and 34 non-sensitizers. Simultaneously, to improve its performance, we changed the incubation time from 5 h to 16 h, deleted the criterion regarding the effects of N-acetylcysteine, and set an exclusion criterion for detergents. These modifications significantly improved its performance. In addition, we examined the following three criteria to judge chemicals as sensitizers: Criterion 1: Fold induction of SLO luciferase activity (FlnSLO-LA)⩾1.4, Criterion 2: the lower limit of the 95% confidence interval of FInSLO-LA⩾1.0, Criterion 3: the intersection of criteria 1 and 2. Among them, Criterion 1 produced the best performance, demonstrating that the accuracy, sensitivity and specificity were 81%, 79%, and 90%, respectively. In addition, we found that the IL-8 Luc assay solubilizing chemicals with X-VIVO substantially improved its performance. Finally, the IL-8 Luc assay combined with DPRA and DEREK could improve substantially its performance. These data suggest that the IL-8 Luc assay is a promising test method to screen skin sensitizers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®