Merck
  • PRMT1 promotes glucose toxicity-induced β cell dysfunction by regulating the nucleo-cytoplasmic trafficking of PDX-1 in a FOXO1-dependent manner in INS-1 cells.
PRMT1 promotes glucose toxicity-induced β cell dysfunction by regulating the nucleo-cytoplasmic trafficking of PDX-1 in a FOXO1-dependent manner in INS-1 cells.
Endocrine (2015-04-16)
Lixia Lv, Hewen Chen, Jiaying Sun, Di Lu, Chen Chen, Dongfang Liu
ABSTRAKT
Protein N-arginine methyltransferase-1 (PRMT1), the major asymmetric arginine methyltransferase, plays important roles in various cellular processes. Previous reports have demonstrated that levels and activities of PRMT1 can vary in animals with type 2 diabetes mellitus. The aim of this study was to assess the expression and mechanism of action of PRMT1 during glucose toxicity-induced β cell dysfunction. Liposome-mediated gene transfection was used to transfect INS-1 cells with siPRMT1, which inhibits PRMT1 expression, and pALTER-FOXO1, which overexpresses forkhead box protein O1 (FOXO1). The cells were then cultured in media containing 5.6 or 25 mmol/L glucose with or without the small molecule PRMT1 inhibitor AMI-1 for 48 h. The protein levels of PRMT1, the arginine methylated protein α-metR, FOXO1, Phospho-FOXO1, pancreas duodenum homeobox-1 (PDX-1), and the intracellular localization of PDX-1 and FOXO1 were then measured by western blotting. FOXO1 methylation was detected by immunoprecipitated with anti-PRMT1 antibody and were immunoblotted with α-metR. The levels of insulin mRNA were measured by real-time fluorescence quantitative PCR. Glucose-stimulated insulin secretion (GSIS) and intracellular insulin content were measured using radioimmunoassays. Intracellular Ca(2+) ([Ca(2+)]i) was detected using Fura-2 AM. Intracellular cAMP levels were measured using ELISA. Chronic exposure to high glucose impaired insulin secretion, decreased insulin mRNA levels and insulin content, increased intracellular [Ca(2+)]i and cAMP levels, and abolishes their responses to glucose. Inhibiting PRMT1 expression improved insulin secretion, increased mRNA levels and insulin content by regulating the intracellular translocation of PDX-1 and FOXO1, decreasing the methylation of FOXO1, and reducing intracellular [Ca(2+)]i and cAMP concentrations. Transient overexpression of constitutively active FOXO1 in nuclear reversed the AMI-1-induced improvement of β cell function without changing arginine methylation. It is concluded therefore that PRMT1 regulates GSIS in INS-1 cells, through enhanced methylation-induced nuclear localization of FOXO1, which subsequently suppresses the nuclear localization of PDX-1. Our results suggest a novel mechanism that might contribute to the deficient insulin secretion observed under conditions of chronically hyperglycemia.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder