Merck
A multi-omic analysis of human naïve CD4+ T cells.
BMC systems biology (2015-11-07)
Christopher J Mitchell, Derese Getnet, Min-Sik Kim, Srikanth S Manda, Praveen Kumar, Tai-Chung Huang, Sneha M Pinto, Raja Sekhar Nirujogi, Mio Iwasaki, Patrick G Shaw, Xinyan Wu, Jun Zhong, Raghothama Chaerkady, Arivusudar Marimuthu, Babylakshmi Muthusamy, Nandini A Sahasrabuddhe, Rajesh Raju, Caitlyn Bowman, Ludmila Danilova, Jevon Cutler, Dhanashree S Kelkar, Charles G Drake, T S Keshava Prasad, Luigi Marchionni, Peter N Murakami, Alan F Scott, Leming Shi, Jean Thierry-Mieg, Danielle Thierry-Mieg, Rafael Irizarry, Leslie Cope, Yasushi Ishihama, Charles Wang, Harsha Gowda, Akhilesh Pandey
ABSTRAKT
Cellular function and diversity are orchestrated by complex interactions of fundamental biomolecules including DNA, RNA and proteins. Technological advances in genomics, epigenomics, transcriptomics and proteomics have enabled massively parallel and unbiased measurements. Such high-throughput technologies have been extensively used to carry out broad, unbiased studies, particularly in the context of human diseases. Nevertheless, a unified analysis of the genome, epigenome, transcriptome and proteome of a single human cell type to obtain a coherent view of the complex interplay between various biomolecules has not yet been undertaken. Here, we report the first multi-omic analysis of human primary naïve CD4+ T cells isolated from a single individual. Integrating multi-omics datasets allowed us to investigate genome-wide methylation and its effect on mRNA/protein expression patterns, extent of RNA editing under normal physiological conditions and allele specific expression in naïve CD4+ T cells. In addition, we carried out a multi-omic comparative analysis of naïve with primary resting memory CD4+ T cells to identify molecular changes underlying T cell differentiation. This analysis provided mechanistic insights into how several molecules involved in T cell receptor signaling are regulated at the DNA, RNA and protein levels. Phosphoproteomics revealed downstream signaling events that regulate these two cellular states. Availability of multi-omics data from an identical genetic background also allowed us to employ novel proteogenomics approaches to identify individual-specific variants and putative novel protein coding regions in the human genome. We utilized multiple high-throughput technologies to derive a comprehensive profile of two primary human cell types, naïve CD4+ T cells and memory CD4+ T cells, from a single donor. Through vertical as well as horizontal integration of whole genome sequencing, methylation arrays, RNA-Seq, miRNA-Seq, proteomics, and phosphoproteomics, we derived an integrated and comparative map of these two closely related immune cells and identified potential molecular effectors of immune cell differentiation following antigen encounter.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, ≥98%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.