Merck
AKT mediated glycolytic shift regulates autophagy in classically activated macrophages.
The international journal of biochemistry & cell biology (2015-07-30)
Sumit Kumar Matta, Dhiraj Kumar
ABSTRAKT
Autophagy is considered as an innate defense mechanism primarily due to its role in the targeting of intracellular pathogens for lysosomal degradation. Here we report inhibition of autophagy as an adaptive response in classically activated macrophages that helps achieve high cellular ROS production and cell death-another hallmark of innate mechanisms. We show prolonged classical activation of Raw 264.7 macrophages by treating them with IFN-γ and LPS inhibited autophagy. The inhibition of autophagy was dependent on nitric oxide (NO) production which activated the AKT-mTOR signaling, the known negative regulators of autophagy. Autophagy inhibition in these cells was accompanied with a shift to aerobic glycolysis along with a decline in the mitochondrial membrane potential (MOMP). The decline in MOMP coupled with autophagy inhibition led to increased mitochondrial content and considerably elevated cellular ROS, eventually causing cell death. Next, using specific siRNA mediated knockdowns we show AKT was responsible for the glycolytic shift and autophagy inhibition in activated macrophages. Surprisingly, AKT knockdown in activated macrophages also rescued them from cell death. Finally we show that AKT mediated autophagy inhibition in the activated macrophages correlated with the depletion of glucose from the extracellular medium, and glucose supplementation not only rescued autophagy levels and reversed other phenotypes of activated macrophages, but also inhibited cell death. Thus we report here a novel link between AKT mediated glycolytic metabolism and autophagy in the activated macrophages, and provide a possible mechanism for sustained macrophage activation in vivo.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder