Merck
Influence of Rictor and Raptor Expression of mTOR Signaling on Long-Term Outcomes of Patients with Hepatocellular Carcinoma.
Digestive diseases and sciences (2014-11-06)
Masaki Kaibori, Nobuaki Shikata, Tatsuma Sakaguchi, Morihiko Ishizaki, Kosuke Matsui, Hiroya Iida, Yoshito Tanaka, Hirokazu Miki, Richi Nakatake, Tadayoshi Okumura, Katsuji Tokuhara, Kentaro Inoue, Jouji Wada, Michio Oda, Mikio Nishizawa, Masanori Kon
ABSTRAKT
Aberrant signaling mediated by the mammalian target of rapamycin (mTOR) occurs at high frequency in hepatocellular carcinoma (HCC), indicating that mTOR is a candidate for targeted therapy. mTOR forms two complexes called mTORC1 (mTOR complexed with raptor) and mTORC2 (mTOR complexed with rictor). There are minor studies of the expression kinetics of mTORC1 and mTORC2 in HCC. We studied 62 patients with HCC who underwent curative resection. We used univariate and multivariate analyses to identify factors that potentially influence disease and overall survival after hepatectomy. The mRNA and protein levels of mTOR, rictor and raptor in cancer and non-cancer tissues were analyzed using quantitative RT-PCR, immunohistochemistry and Western blotting. High ratio of the levels of rictor and raptor mRNAs in tumors was identified as independent prognostic indicators for disease-free survival. Low and high levels of preoperative serum albumin and mTOR mRNA in the tumor, respectively, were identified as independent indicators of overall survival. HCC is likely to recur early after hepatic resection in patients with high levels of mTOR and rictor mRNAs and high rictor/raptor ratios in cancer tissues. We conclude that analysis of mTOR expression in cancer tissues represents an essential strategy to predict HCC recurrence after curative treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture