Merck
  • Glycyrrhetinic Acid protects the heart from ischemia/reperfusion injury by attenuating the susceptibility and incidence of fatal ventricular arrhythmia during the reperfusion period in the rat hearts.
Glycyrrhetinic Acid protects the heart from ischemia/reperfusion injury by attenuating the susceptibility and incidence of fatal ventricular arrhythmia during the reperfusion period in the rat hearts.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-05-30)
Hong-Jin Wu, Ji-Yuan Yang, Min Jin, Sheng-Qi Wang, De-Lin Wu, Yu-Na Liu, Xu Yan, Cui Yang, Ge Zhang, Jing He
ABSTRAKT
Licorice has been used to treat many diseases, including palpitations, in both Eastern and Western societies for thousands of years. It has been reported that glycyrrhetinic acid (GA), an aglycone saponin extracted from licorice root, exerts protective effects on the cardiovascular system, limits infarct sizes and protects against the development of arrhythmia. However, the mechanisms underlying the effects of glycyrrhetinic acid on the cardiovascular system remain poorly understood. This study aimed to determine the mechanisms underlying the protective effects of GA against lethal cardiac arrhythmias induced via ischemia-reperfusion in rat hearts, and to examine its electropharmacological properties. Anesthetized rats were divided into control (CTL), GA5, GA10, and GA20 groups. GA was administered intravenously 15 min before the occlusion of the left anterior descending coronary artery, at dosages of 5, 10 and 20 mg/kg, respectively. Single ventricular myocytes were isolated using enzymolysis. The whole-cell patch clamp technique was utilized to record Ica, L, Ito and action potentials (APs). During reperfusion, the incidence of ventricular fibrillation (VF) was decreased in each of the groups compared with the CTL group (p<0.05). The ventricular tachycardia (VT)/VF score was significantly decreased in the GA20 group. Action potential durations (APDs) were prolonged by GA; both L-type calcium current (Ica-L) and transient outward potassium current (Ito) were blocked in a concentration-dependent manner by GA. These results suggest that GA attenuates both the susceptibility to and the incidence of fatal ventricular arrhythmia during reperfusion in rat hearts via the prolongation of the APD and the inhibition of both Ica-L and Ito. GA appears to be a promising antiarrhythmic agent in the setting of ischemia/reperfusion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)