Merck
Microbial colonization affects the efficiency of photovoltaic panels in a tropical environment.
Journal of environmental management (2015-04-25)
Marcia A Shirakawa, Roberto Zilles, Andre Mocelin, Christine C Gaylarde, Anna Gorbushina, Gabriele Heidrich, Mauro C Giudice, Gilda M B Del Negro, Vanderley M John
ABSTRAKT
Sub-aerial biofilm (SAB) development on solar panels was studied in São Paulo. After 6, 12 and 18 months' exposure, photovoltaic panels were covered by increasing proportions of organic matter (42%, 53% and 58%, respectively). Fungi were an important component of these biofilms; very few phototrophs were found. Major microorganisms detected were melanised meristematic ascomycetes and pigmented bacterial genera Arthrobacter and Tetracoccus. While diverse algae, cyanobacteria and bacteria were identified in biofilms at 6 and 12 months, diversity at a later stage was reduced to that typical for SAB: the only fungal group detected in 18 month biofilm was the meristematic Dothideomycetes and the only phototrophs Ulothrix and Chlorella. Photovoltaic modules showed significant power reductions after 6, 12 (both 7%) and 18 (11%) months. The lack of difference in power reduction between 6 and 12 months reflects the dual nature of soiling, which can result from the deposition of particulates as well as from SAB fouling. Although 12-month old SAB demonstrated an almost 10-fold increase in fungal colonization and a higher organic content, the larger non-microbial particles (above 10 μm), which were important for efficiency reduction of lightly-biofilmed panels, were removed by high rainfall just before the 12-month sampling.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis