MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Design, synthesis and pharmacobiological evaluation of novel acrylic acid derivatives acting as lipoxygenase and cyclooxygenase-1 inhibitors with antioxidant and anti-inflammatory activities.
Design, synthesis and pharmacobiological evaluation of novel acrylic acid derivatives acting as lipoxygenase and cyclooxygenase-1 inhibitors with antioxidant and anti-inflammatory activities.
European journal of medicinal chemistry (2010-11-26)
Eleni Pontiki, Dimitra Hadjipavlou-Litina, Konstantinos Litinas, Orazio Nicolotti, Angelo Carotti
ABSTRACT
A series of novel acrylic acid derivatives bearing at the 3 position thienyl, furfuryl and 3,5-ditert-butyl-4-hydroxyphenyl substituents have been designed, synthesized and tested as potential dual lipoxygenase/cyclooxygenase-1 (LOX/COX-1) inhibitors and as antioxidant and anti-inflammatory agents. Some compounds have shown moderate antioxidant and COX-1 inhibitory activities, very good anti-inflammatory activity and an inhibition of soybean lipoxygenase (LOX) higher than caffeic acid. In particular, compound 4I disclosed a moderate in vitro LOX inhibition with an IC(50) = 100 μM whereas compounds 1I and 2II exhibited the best, albeit poor, activity as COX-1 inhibition (75% inhibition at 100 μM). Good radical scavenging properties were shown by compounds 4I, 3I and 1II. Docking simulations performed on LOX inhibitor 4I and COX-1 inhibitor 1I indicated that hydrophobic key interactions may govern the enzyme-inhibitor binding.
Sigma-Aldrich
Nordihydroguaiaretic acid, ≥97.0% (HPLC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)