MilliporeSigma
Quantitative PCR for genetic markers of human fecal pollution.
Applied and environmental microbiology (2009-07-14)
Orin C Shanks, Catherine A Kelty, Mano Sivaganesan, Manju Varma, Richard A Haugland
ABSTRACT
Assessment of health risk and fecal bacterial loads associated with human fecal pollution requires reliable host-specific analytical methods and a rapid quantification approach. We report the development of quantitative PCR assays for quantification of two recently described human-specific genetic markers targeting Bacteroidales-like cell surface-associated genes. Each assay exhibited a range of quantification from 10 to 1 x 10(6) copies of target DNA. For each assay, internal amplification controls were developed to detect the presence or absence of amplification inhibitors. The assays predominantly detected human fecal specimens and exhibited specificity levels greater than 97% when tested against 265 fecal DNA extracts from 22 different animal species. The abundance of each human-specific genetic marker in primary effluent wastewater samples collected from 20 geographically distinct locations was measured and compared to quantities estimated by real-time PCR assays specific for rRNA gene sequences from total Bacteroidales and enterococcal fecal microorganisms. Assay performances combined with the prevalence of DNA targets in sewage samples provide experimental evidence supporting the potential application of these quantitative methods for monitoring fecal pollution in ambient environmental waters.
Sigma-Aldrich
Phosphate buffered saline, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, Autoclaved, pH 7.2 (25 °C)
Sigma-Aldrich
Phosphate buffered saline, pH 7.4, contains BSA, powder
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Phosphate buffered saline, pH 7.6 (25 °C)
Sigma-Aldrich
Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, with 5% nonfat milk, powder (dry milled), pH 7.3
Sigma-Aldrich
Phosphate buffered saline, powder, pH 7.4, for preparing 5 L solutions
Sigma-Aldrich
Phosphate Buffered Saline with 0.05% TWEEN® 20, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Phosphate buffered saline, BioUltra, solution
Sigma-Aldrich
Phosphate buffered saline, BioUltra, pH 7.4 ( in solution), contains TWEEN 20, tablet
Sigma-Aldrich
Phosphate buffered saline, tablet
Sigma-Aldrich
Phosphate buffered saline, BioUltra, washing buffer for peroxidase conjugates in Western Blotting, 10x concentrate
Sigma-Aldrich
Phosphate Buffered Saline, 10× PBS for Western blots and IP
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Bovine Serum Albumin, dimer mol wt ~132 kDa
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated, Molecular Biology
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture