MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibitory and bactericidal activity of selected South African honeys and their solvent extracts against clinical isolates of Helicobacter pylori.
Inhibitory and bactericidal activity of selected South African honeys and their solvent extracts against clinical isolates of Helicobacter pylori.
Pakistan journal of pharmaceutical sciences (2013-09-17)
Christy Echakachi Manyi-Loh, Anna Maria Clarke, Ezekiel Green, Roland Ndip Ndip
ABSTRACT
The growing problem of antibiotic resistance by Helicobacter pylori demands the search for novel compounds, especially from natural sources. We evaluated the anti-H. pylori activity of six local honeys at different concentrations as well as their solvent extracts by the Hole Plate diffusion method. The minimum inhibitory concentration (MIC50) of the two most active extracts of each honey was determined by the broth microdilution method; and the time kill assay of the most active extract of each honey determined by viability studies. Data were analyzed by one-way ANOVA test at 95% significance level. All the honey varieties as well as their solvent extracts demonstrated varying levels of antibacterial activity based on different mean zone diameters [16.0mm (crude) to 22.2mm (extract)] and percentage susceptibilities [73.3% (crude) to 93.3% (extract)] of the test isolates. The chloroform extracts of Pure Honey (PH) and Champagne Royal Train (CRT) recorded MIC50 ranges of 0.01-10% and 0.625-10 % (v/v) respectively; that were not significantly different (P > 0.05) from amoxicillin (0.001-1.25mg/mL), the positive control. The most potent bactericidal effect against the test isolates was obtained with 5% v/v (1/2 MIC) concentration of chloroform extract of PH from 42-72 hrs. In conclusion, these honeys and their extracts could be leads for further investigation in the discovery of new natural anti-H. pylori compounds.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, ≥99.5%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Diethyl ether, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Diethyl ether, puriss., contains ~5 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer, meets analytical specification of Ph. Eur., BP, ≥99.5% (GC)
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor