MilliporeSigma
  • Treatment of acquired muscle spasticity using phenol peripheral nerve blocks.
Treatment of acquired muscle spasticity using phenol peripheral nerve blocks.
Orthopedics (1995-02-01)
M J Botte, R A Abrams, S C Bodine-Fowler
ABSTRACT
The use of phenol motor nerve blocks is advantageous in the early period of acquired spasticity (ie, that occurring following traumatic brain injury or incomplete spinal cord injury), when increased muscle tone is often the most severe. Because acquired spasticity is dynamic and usually improves slowly, a temporary treatment method used to ameliorate increased muscle tone is desirable. Phenol nerve infiltration provides a temporary motor nerve block that lasts for weeks or months. It allows passive limb mobilization in a comprehensive rehabilitation program that attempts to prevent fixed soft tissue contractures. Permanent or irreversible methods such as operative tendon lengthening, muscle release or recession, or neurectomy are usually best delayed until the spasticity has become static, when the need for surgical correction becomes more firmly indicated, and outcomes of operative intervention are more predictable. Although phenol nerve blocks were initially administered at the spinal cord level to control spasticity, the potential side effects have caused a loss of popularity of this method of administration. The safer and more common use of phenol infiltration at the peripheral nerve level is now more accepted for brain injury and spinal cord injury patients. This report reviews the indications, current concepts, and development of the different methods used to administer phenol nerve blocks. Comparisons to other methods to control spasticity are discussed.
Sigma-Aldrich
Phenol, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, SAJ first grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Supelco
Phenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Pricing and availability is not currently available.
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Pricing and availability is not currently available.
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol, natural, 97%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Pandy′s reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Phenol stock solution, 100 mg/dL, standard
Pricing and availability is not currently available.