MilliporeSigma
  • The effect of oxidized cholesterol on barrier functions and IL-10 mRNA expression in human intestinal epithelium co-cultured with dendritic cells in the transwell system.
The effect of oxidized cholesterol on barrier functions and IL-10 mRNA expression in human intestinal epithelium co-cultured with dendritic cells in the transwell system.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-04-15)
Maciej Chalubinski, Katarzyna Wojdan, Paulina Gorzelak, Maciej Borowiec, Marlena Broncel
ABSTRACT
The intestinal epithelium is exposed to oxygenated cholesterol products present in foodstuffs. In vitro studies demonstrate the effect of oxysterols on cytokine release by intestinal cells cultured alone. However, physiologically, the response of the intestinal epithelium to external agents occurs in the presence of dendritic cells (DCs). The aim of the study was to analyze the effect of 7-ketocholesterol on the barrier functions and IL-10 mRNA expression of Caco-2 cells in the presence of DCs, and secondly, on IL-10 mRNA expression in DCs. Caco-2 cells were co-cultured with monocyte-derived dendritic cells and induced with 7-ketocholesterol in a transwell system. DCs did not affect the transepithelial electrical resistance (TER) of the Caco-2 cell monolayer, but increased IL-10 mRNA expression in Caco-2 cells. 7-ketocholesterol decreased the TER of Caco-2 cells co-cultured with DCs and diminished IL-10 mRNA expression in Caco-2 cells induced by the presence of DCs. IL-10 mRNA expression fell in DCs co-cultured with Caco-2 cells after treatment with 7-ketocholesterol. Oxidized cholesterols present in gut mucosa may contribute to the decrease of epithelial barrier functions and the inappropriate development of an inflammatory response to food compounds.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Glutamine–Penicillin–Streptomycin solution, L-glutamine 200 mM, streptomycin 10 mg/mL, penicillin 10,000 units, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
5-Cholesten-3β-ol-7-one, ≥90%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, Alpha Modification, with sodium bicarbonate, without L-glutamine, ribonucleosides and deoxyribonucleosides, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland