MilliporeSigma
  • The inhibition of tyrosine kinase receptor signalling in leiomyosarcoma cells using the small molecule kinase inhibitor PTK787/ZK222584 (Vatalanib®).
The inhibition of tyrosine kinase receptor signalling in leiomyosarcoma cells using the small molecule kinase inhibitor PTK787/ZK222584 (Vatalanib®).
International journal of oncology (2014-10-24)
Andreas K A Gaumann, Hannes C A Drexler, Sven A Lang, Oliver Stoeltzing, Simone Diermeier-Daucher, Elisabeth Buchdunger, Jeanette Wood, Guido Bold, Georg Breier
ABSTRACT
Leiomyosarcomas remain challenging tumors to manage and novel therapy strategies besides radiation and conventional chemotherapy are needed. Targeting angiogenesis by inhibition of vascular endothelial growth factor (VEGF) receptor tyrosine kinases (RTKs) of the tumor vasculature with small molecules is a promising new therapy. It has been shown recently that these receptors are not only expressed on tumor endothelium but also on tumor cells themselves. Thus, we investigated the expression of members of the VEGF receptor (VEGFR) family and corresponding growth factors in leiomyosarcoma tissue specimens and in the leiomyosarcoma cell lines SK-LMS-1 and SK-UT-1. We evaluated the influence of the VEGFR inhibitor PTK787/ZK222584 (PTK787) on cell growth, migration, apoptosis and phosphorylation of intracellular signalling molecules. In human leiomyosarcoma tissue specimens VEGFR‑1/-2 and platelet-derived growth factor receptor (PDGFR-β) were strongly expressed. Both leiomyosarcoma cell lines expressed VEGFR‑1/-3 and PDGFR-β but VEGFR-2 protein expression was positive only in SK-UT-1. SK-LMS-1 and SK-UT-1 cells secreted high and low amounts of VEGF-A, respectively, whereas PDGF-BB secretion was similar in both cell lines. Application of PTK787 led to partial inhibition of PDGF-BB-activated AKT/p90RSK and ERK1/2 signalling pathways. In contrast, protein phosphorylation was not affected by PTK787 in VEGF-A-treated cells. PTK787 turned out to inhibit cell migration even though no effects were observed upon stimulation with VEGF-A or PDGF-BB. In line, cell growth in leiomyosarcoma cell lines remained unchanged upon PTK787 treatment alone and with subsequent VEGF-A- or PDGF-BB-stimulation. However, VEGF-A, but not PDGF-BB-treated cells showed increased cell death upon PTK787 treatment. VEGFR family members are expressed in leiomyosarcomas in vivo and in vitro. Upon receptor stimulation, PTK787 is able to inhibit subsequent phosphorylation events and influences cell survival but not metabolic activity and migration. Thus, the inhibitor is possibly an additional option in the treatment of leiomyosarcomas.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
IgG1 Isotype Control from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Ethanol, 94.8-95.8%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%