MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A novel and efficient nicotine vaccine using nano-lipoplex as a delivery vehicle.
A novel and efficient nicotine vaccine using nano-lipoplex as a delivery vehicle.
Human vaccines & immunotherapeutics (2013-10-05)
Yun Hu, Hong Zheng, Wei Huang, Chenming Zhang
ABSTRACT
A number of vaccines conjugating nicotine haptens with carrier proteins have been developed to combat nicotine caused tobacco dependence. Some vaccines, such as NicVAX, NicQb, advanced into clinical trials, but none of them were successful. Most of those vaccines have some innate disadvantages such as low nicotine loading capacity, easy degradation, and vulnerable to the clearance by reticulo-endothelial system (RES). Thus, there is undoubtedly an urgent need for developing novel vaccines against nicotine addiction. In this study, we assembled a liposome-protein based nanoparticle as a nicotine hapten delivery system. The nanoparticle (Scheme 1) was constructed by conjugating a model hapten carrier protein, bovine serum albumin (BSA), to cationic liposomes. This nano-sized complex, lipoplex, was characterized using zetasizer, transmission electron microscope (TEM), and flow cytometry. The efficacy of the lipoplex vaccine was evaluated in mice and compared with that of Nicotine-BSA conjugate (Nic-BSA). The lipoplex vaccine with Alum was able to elicit the highest NicAb titer of 11169±2112, which was significantly higher than that induced by either the vaccine without Alum or Nic-BSA with Alum. The significant immunostimulatory effect of this nano-lipoplex may provide a novel strategy to improve the immunogenic ability of current nicotine vaccines or other vaccines using small molecules as immunogens.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%