MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Autophagy-mediated stress response in motor neurons after hypothermic spinal cord ischemia in rabbits.
Autophagy-mediated stress response in motor neurons after hypothermic spinal cord ischemia in rabbits.
Journal of vascular surgery (2014-05-14)
Satoshi Fujita, Masahiro Sakurai, Hironori Baba, Koji Abe, Ryuji Tominaga
ABSTRACT
The development of spinal cord injury is believed to be related to the vulnerability of spinal motor neurons to ischemia. However, the mechanisms underlying this vulnerability have not been fully investigated. Previously, we reported that spinal motor neurons are lost likely due to autophagy and that local hypothermia prevents such spinal motor neuron death. Therefore, we investigated the role of autophagy in normothermic and hypothermic spinal cord ischemia using an immunohistochemical analysis of Beclin 1 (BCLN1; B-cell leukemia 2 protein [Bcl-2] interacting protein), Bcl-2, and γ-aminobutyric acid type-A receptor-associated protein (GABARAP), which are considered autophagy-related proteins. We used rabbit normothermic and hypothermic transient spinal cord ischemia models using a balloon catheter. Neurologic function was assessed according to the Johnson score, and the spinal cord was removed at 8 hours and 1, 2, and 7 days after reperfusion, and morphologic changes were examined using hematoxylin and eosin staining. A Western blot analysis and histochemical study of BCLN1, Bcl-2, and GABARAP, and double-labeled fluorescent immunocytochemical studies were performed. There were significant differences in the physiologic function between the normothermic model and hypothermic model after the procedure (P < .05). In the normothermic model, most of the motor neurons were selectively lost at 7 days of reperfusion (P < .001 compared with the sham group), and they were preserved in the hypothermic model (P = .574 compared with the sham group). The Western blot analysis revealed that the sustained expression of the autophagy markers, BCLN1 and GABARAP, was observed (P < .001 compared with the sham group) and was associated with neuronal cell death in normothermic ischemic conditions. In hypothermic ischemic conditions, the autophagy inhibitory protein Bcl-2 was powerfully induced (P < .001 compared with the sham group) and was associated with blunted expression of BCLN1 and GABARAP and neuronal cell survival. The double-label fluorescent immunocytochemical study revealed that immunoreactivitiy for BCLN1, Bcl-2, and GABARAP was induced in the same motor neurons. These data suggest that the prolonged induction of autophagy might be a potential factor responsible for delayed motor neuron death, and the induction of the autophagy inhibitory protein Bcl-2 using hypothermia might limit autophagy and protect against delayed motor neuron death.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Glycine
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis