MilliporeSigma
  • In vivo hepatogenic capacity and therapeutic potential of stem cells from human exfoliated deciduous teeth in liver fibrosis in mice.
In vivo hepatogenic capacity and therapeutic potential of stem cells from human exfoliated deciduous teeth in liver fibrosis in mice.
Stem cell research & therapy (2015-09-12)
Takayoshi Yamaza, Fatima Safira Alatas, Ratih Yuniartha, Haruyoshi Yamaza, Junko K Fujiyoshi, Yusuke Yanagi, Koichiro Yoshimaru, Makoto Hayashida, Toshiharu Matsuura, Reona Aijima, Kenji Ihara, Shouichi Ohga, Songtao Shi, Kazuaki Nonaka, Tomoaki Taguchi
ABSTRACT
Liver transplantation is a gold standard treatment for intractable liver diseases. Because of the shortage of donor organs, alternative therapies have been required. Due to their potential to differentiate into a variety of mature cells, stem cells are considered feasible cell sources for liver regeneration. Stem cells from human exfoliated deciduous teeth (SHED) exhibit hepatogenic capability in vitro. In this study, we investigated their in vivo capabilities of homing and hepatocyte differentiation and therapeutic efficacy for liver disorders in carbon tetrachloride (CCl4)-induced liver fibrosis model mice. We transplanted SHED into CCl4-induced liver fibrosis model mice through the spleen, and analyzed the in vivo homing and therapeutic effects by optical, biochemical, histological, immunological and molecular biological assays. We then sorted human leukocyte antigen-ABC (HLA-ABC)-positive cells from primary CCl4-damaged recipient livers, and analyzed their fusogenicity and hepatic characteristics by flow cytometric, genomic DNA, hepatocyte-specific gene assays. Furthermore, we examined the treatment effects of HLA-positive cells to a hepatic dysfunction by a secondary transplantation into CCl4-treated mice. Transplanted SHED homed to recipient livers, and expressed HLA-ABC, human hepatocyte specific antigen hepatocyte paraffin 1 and human albumin. SHED transplantation markedly recovered liver dysfunction and led to anti-fibrotic and anti-inflammatory effects in the recipient livers. SHED-derived HLA-ABC-positive cells that were sorted from the primary recipient liver tissues with CCl4 damage did not fuse with the host mouse liver cells. Sorted HLA-positive cells not only expressed human hepatocyte-specific genes including albumin, cytochrome P450 1A1, fumarylacetoacetase, tyrosine aminotransferase, uridine 5'-diphospho-glucuronosyltransferase, transferrin and transthyretin, but also secreted human albumin, urea and blood urea nitrogen. Furthermore, SHED-derived HLA-ABC-positive cells were secondary transplanted into CCl4-treated mice. The donor cells homed into secondary recipient livers, and expressed hepatocyte paraffin 1 and human albumin, as well as HLA-ABC. The secondary transplantation recovered a liver dysfunction in secondary recipients. This study indicates that transplanted SHED improve hepatic dysfunction and directly transform into hepatocytes without cell fusion in CCl4-treated mice, suggesting that SHED may provide a feasible cell source for liver regeneration.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%