MilliporeSigma
DMSO induces dehydration near lipid membrane surfaces.
Biophysical journal (2015-07-23)
Chi-Yuan Cheng, Jinsuk Song, Jolien Pas, Lenny H H Meijer, Songi Han
ABSTRACT
Dimethyl sulfoxide (DMSO) has been broadly used in biology as a cosolvent, a cryoprotectant, and an enhancer of membrane permeability, leading to the general assumption that DMSO-induced structural changes in cell membranes and their hydration water play important functional roles. Although the effects of DMSO on the membrane structure and the headgroup dehydration have been extensively studied, the mechanism by which DMSO invokes its effect on lipid membranes and the direct role of water in this process are unresolved. By directly probing the translational water diffusivity near unconfined lipid vesicle surfaces, the lipid headgroup mobility, and the repeat distances in multilamellar vesicles, we found that DMSO exclusively weakens the surface water network near the lipid membrane at a bulk DMSO mole fraction (XDMSO) of <0.1, regardless of the lipid composition and the lipid phase. Specifically, DMSO was found to effectively destabilize the hydration water structure at the lipid membrane surface at XDMSO <0.1, lower the energetic barrier to dehydrate this surface water, whose displacement otherwise requires a higher activation energy, consequently yielding compressed interbilayer distances in multilamellar vesicles at equilibrium with unaltered bilayer thicknesses. At XDMSO >0.1, DMSO enters the lipid interface and restricts the lipid headgroup motion. We postulate that DMSO acts as an efficient cryoprotectant even at low concentrations by exclusively disrupting the water network near the lipid membrane surface, weakening the cohesion between water and adhesion of water to the lipid headgroups, and so mitigating the stress induced by the volume change of water during freeze-thaw.
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Pricing and availability is not currently available.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Pricing and availability is not currently available.