MilliporeSigma
  • Comparative analysis of goitrogenic effects of phenylthiourea and methimazole in zebrafish embryos.
Comparative analysis of goitrogenic effects of phenylthiourea and methimazole in zebrafish embryos.
Reproductive toxicology (Elmsford, N.Y.) (2015-05-13)
Eva Fetter, Lisa Baldauf, Dillon F Da Fonte, Julia Ortmann, Stefan Scholz
ABSTRACT
Craniofacial malformations, reduced locomotion and induction of genes encoding for enzymes involved in thyroid hormone synthesis were assessed using methimazole and N-phenylthiourea in zebrafish embryos. Gene expression, the most sensitive endpoint (EC50_MMI=372-765μM, EC50_PTU=7.6-8.6μM), was analysed in wild-type and in a transgenic strain, tg(tg:mCherry), expressing mCherry fluorescence protein under the control of the thyroglobulin gene. Reduction of locomotion and craniofacial malformations were observed at one or two orders of magnitude above concentrations affecting gene expression, respectively. Both effects could be linked to the malformations caused by reduced thyroxin levels. Our results show that due to the presence of the autoregulatory loop of the hypothalamus-pituitary-thyroid axis, various molecular initiating events of thyroid disruption are amenable for the zebrafish embryo. We propose the tg(tg:mCherry) bioassay as a sensitive tool in medium scale screening of goitrogens, given the minimal effort for sample preparation and analysis of gene expression.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Supelco
L-Thyroxine (T4) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
5-Propyl-2-thiouracil, ≥98%
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
N-Phenylthiourea, ≥98%