MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Understanding the regulation of pituitary progesterone receptor expression and phosphorylation.
Understanding the regulation of pituitary progesterone receptor expression and phosphorylation.
Reproduction (Cambridge, England) (2015-05-01)
Ana Gordon, José C Garrido-Gracia, Rafaela Aguilar, José E Sánchez-Criado
ABSTRACT
Administration of human FSH (hFSH) during the diestrus phase in cyclic rats is followed by a reduction in the preovulatory LH surge. This inhibitory action of FSH involves a decrease in the stimulatory effect of gonadotrope progesterone receptor (PR) activation, in a ligand-dependent (progesterone) and -independent (GNRH) manner. PR activation and action are mandatory for LH surge, and are dependent on the phosphorylation of serine (Ser) residues. Together with this post-translational modification, PR is marked for downregulation by proteasome machinery. These experiments used the western blotting technique to measure pituitary expression of PR-A and PR-B isoforms and phosphorylation levels of Ser294 and Ser400 PR-B in rats bearing i) hFSH treatment or ii) PR downregulation. Treatment with hFSH reduced LH secretion and increased that of estradiol in proestrus afternoon. hFSH injections, without altering PR-A and PR-B content or ratio, caused a reduction in phosphorylation of Ser294 and Ser400 but only when pituitaries were previously challenged with progesterone or GNRH for 2 h. However, while pSer294 levels increased after 2 h of pituitary incubation with progesterone or GNRH, those of pSer400 were not modified by these in vitro treatments. Finally, progesterone had a biphasic effect: in 2-h incubations increased pituitary PR-A and PR-B content, but after 8 h caused downregulation and altered PR-A:PR-B ratio. The results provide a potential mechanism through which LH levels are decreased by hFSH administration and better understanding of the control of PR expression and phosphorylation in rat pituitaries.
Sigma-Aldrich
Phenol Red, ACS reagent
SAFC
L-Glutamine
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))