MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Maintenance of HL-1 cardiomyocyte functional activity in PEGylated fibrin gels.
Maintenance of HL-1 cardiomyocyte functional activity in PEGylated fibrin gels.
Biotechnology and bioengineering (2015-02-07)
Laura R Geuss, Alicia C B Allen, Divya Ramamoorthy, Laura J Suggs
ABSTRACT
Successful cellular cardiomyoplasty is dependent on biocompatible materials that can retain the cells in the myocardium in order to promote host tissue repair following myocardial infarction. A variety of methods have been explored for incorporating a cell-seeded matrix into the heart, the most popular options being direct application of an injectable system or surgical implantation of a patch. Fibrin-based gels are suitable for either of these approaches, as they are biocompatible and have mechanical properties that can be tailored by adjusting the initial fibrinogen concentration. We have previously demonstrated that conjugating amine-reactive homo-bifunctional polyethylene glycol (PEG) to the fibrinogen prior to crosslinking with thrombin can increase stability both in vivo and in vitro. Similarly, when mesenchymal stem cells are combined with PEGylated fibrin and injected into the myocardium, cell retention can be significantly increased and scar tissue reduced following myocardial infarction. We hypothesized that this gel system could similarly promote cardiomyocyte viability and function in vitro, and that optimizing the mechanical properties of the hydrogel would enhance contractility. In this study, we cultured HL-1 cardiomyocytes either on top of plated PEGylated fibrin (2D) or embedded in 3D gels and evaluated cardiomyocyte function by assessing the expression of cardiomyocyte specific markers, sarcomeric α-actin, and connexin 43, as well as contractile activity. We observed that the culture method can drastically affect the functional phenotype of HL-1 cardiomyocytes, and we present data suggesting the potential use of PEGylated fibrin gel layers to prepare a sheet-like construct for myocardial regeneration.
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
Calcium, SAJ first grade, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%