MilliporeSigma
  • Involvement of cAMP-PKA pathway in adenosine A1 and A2A receptor-mediated regulation of acetaldehyde-induced activation of HSCs.
Involvement of cAMP-PKA pathway in adenosine A1 and A2A receptor-mediated regulation of acetaldehyde-induced activation of HSCs.
Biochimie (2015-05-10)
Yaru Yang, He Wang, Xiongwen Lv, Qi Wang, Han Zhao, Feng Yang, Yan Yang, Jun Li
ABSTRACT
The present study was undertaken to investigate the mechanism by which adenosine receptors (ARs)-mediated the cAMP/PKA/CREB signal pathway regulates the activation of acetaldehyde-induced hepatic stellate cells (HSCs). Primary HSCs were isolated from SD rats, cultured in vitro, and activated with different concentrations of acetaldehyde at different time points. Quantitative real-time PCR and Western blotting were used to quantify both protein and mRNA levels of the four AR (A1R, A2AR, A2BR, and A3R) in rat HSCs. Selective inhibitors of PDEs and the Gi/o protein pathway, general AR agonists, and AR subtype specific agents were used to study the AR signaling. The level of cAMP was measured by radio-immunoassay, and the expression of α-SMA, collagen type I and III, PKA and p-CREB were also detected by Western blotting. Acetaldehyde could significantly promote HSC proliferation, with a maximum stimulatory effect observed at 48 h after exposure to 200 μM acetaldehyde. All four AR subtypes could be present in rat HSCs, and the mRNA and protein expression levels for A2AR and A1R in much greater abundance than those for A2BR and A3R. The expression of A2AR and A1R was significantly increased in acetaldehyde-induced HSCs as compared with that of control group, whereas the expression of A2BR and A3R remained unaffected by the addition of acetaldehyde. Curiously, there is coupling of A2AR to the Gs-AC signaling, as well as coupling of A1R to the Gi/o-AC signaling pathway in acetaldehyde-induced HSCs. Both the A2AR and A1R antagonists could suppress the activation of HSC, although they have opposing effects on cAMP signal transduction. These results suggested that a combination of cAMP/PKA/CREB signals via A2AR and A1R likely mediate the activation of acetaldehyde-induced HSCs, and A1R coupled to the Gi/o-AC signaling pathway may be masked by the more predominant A2AR that coupled to the Gs-AC signaling pathway.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
5′-(N-Ethylcarboxamido)adenosine, powder
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent