MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Bioconcentration and Elimination of the Dithiocarbamate Fungicide Polycarbamate in Marine Teleost Fish and Polychaete.
Bioconcentration and Elimination of the Dithiocarbamate Fungicide Polycarbamate in Marine Teleost Fish and Polychaete.
Bulletin of environmental contamination and toxicology (2015-04-24)
Takeshi Hano, Nobuyuki Ohkubo, Kumiko Kono, Hiroyuki Tanaka
ABSTRACT
We evaluated the bioconcentration and elimination of polycarbamate, a popular antifoulant classified as a dithiocarbamate fungicide, in a marine teleost fish, mummichog (Fundulus heteroclitus), and a polychaete (Perinereis nuntia). Following polycarbamate exposure and depuration, the calculated bioconcentration factor (BCF) and elimination rate constant (k2) were 3.1 and 0.17 day(-1) in fish and 1.5-7.8 and 0.13-0.18 day(-1) in the polychaete, indicating that the fungicide has low bioconcentration potential in both organisms. Given the BCF of 3.1, the permissible environmental concentration level of polycarbamate in water was calculated to be 6.1 µg L(-1) to satisfy the threshold pesticide residue levels in fish permitted by law, which is far below the permissible levels in Hiroshima Bay, Japan. Therefore, we consider that the current polycarbamate contamination level in terms of bioconcentration is not likely to be an alarming issue in coastal environments.
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SAFC
L-Cysteine
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
L-Cysteine hydrochloride hydrate, 99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis