MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Endothelial CD36 Contributes to Postischemic Brain Injury by Promoting Neutrophil Activation via CSF3.
Endothelial CD36 Contributes to Postischemic Brain Injury by Promoting Neutrophil Activation via CSF3.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2015-11-06)
Lidia Garcia-Bonilla, Gianfranco Racchumi, Michelle Murphy, Josef Anrather, Costantino Iadecola
ABSTRACT
The scavenger receptor CD36 is a critical factor initiating ischemic brain injury, but the cell type(s) expressing CD36 and responsible for its harmful effects remain unknown. Using bone marrow (BM) chimeras subjected to transient middle cerebral artery occlusion, we found that CD36(-/-) mice transplanted with wild-type (WT) BM (WT→CD36(-/-)) have smaller infarcts (-67%), comparable with those of mice lacking CD36 both in brain and hematogenous cells (CD36(-/-) →CD36(-/-); - 72%). Conversely, WT mice receiving CD36(-/-) BM (CD36(-/-) →WT) have infarcts similar to WT→WT mice, suggesting that CD36 in the host brain (i.e., in microglia and endothelial cells), and not in hematogenous cells is involved in the damage. As anticipated, postischemic neutrophil infiltration in CD36(-/-) →CD36(-/-) mice was attenuated. Surprisingly, however, in WT→CD36(-/-) mice, in which infarcts were small, neutrophil infiltration was large and similar to that of CD36(-/-) →WT mice, in which infarcts were not reduced. Postischemic neutrophil free radical production was attenuated in WT→CD36(-/-) mice compared with CD36(-/-) →WT mice, whereas expression of the neutrophil activator colony-stimulating factor 3 (CSF3) was suppressed in CD36(-/-) cerebral endothelial cells, but not microglia. In CD36(-/-) cerebral endothelial cultures exposed to extracts from stroke brains, the upregulation of CSF3, but not neutrophil attractant chemokines, was suppressed. Intracerebroventricular administration of CSF3, 24 h after stroke, reconstituted neutrophil radical production and increased infarct volume in WT→CD36(-/-) mice. The findings identify endothelial cells as a key player in the deleterious effects of CD36 in stroke, and unveil a novel role of endothelial CD36 in enabling neutrophil neurotoxicity through CSF3. Ischemic stroke is a leading cause of death and disability worldwide with limited therapeutic options. The inflammatory response initiated by cerebral ischemia-reperfusion contributes to ischemic brain injury and is a potential therapeutic target. Here we report that CD36, an innate immunity receptor involved in the initiation of postischemic inflammation, is a previously unrecognized regulator of neutrophil cytotoxicity. The effect is mediated by endothelial CD36 via upregulation of the neutrophil activator CSF3 in cerebral endothelial cells. Therefore, approaches to modulate cerebral endothelial CD36 signaling or to neutralize CSF3 may provide novel therapeutic opportunities to ameliorate postischemic inflammatory injury.
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Dihydroethidium, ≥95%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)