MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Epigenetic silencing of Oct4 by a complex containing SUV39H1 and Oct4 pseudogene lncRNA.
Epigenetic silencing of Oct4 by a complex containing SUV39H1 and Oct4 pseudogene lncRNA.
Nature communications (2015-07-15)
Michele Scarola, Elisa Comisso, Rhena Pascolo, Riccardo Chiaradia, Rosa Maria Marion, Claudio Schneider, Maria A Blasco, Stefan Schoeftner, Roberta Benetti
ABSTRACT
Pseudogene-derived, long non-coding RNAs (lncRNAs) act as epigenetic regulators of gene expression. Here we present a panel of new mouse Oct4 pseudogenes and demonstrate that the X-linked Oct4 pseudogene Oct4P4 critically impacts mouse embryonic stem cells (mESCs) self-renewal. Sense Oct4P4 transcription produces a spliced, nuclear-restricted lncRNA that is efficiently upregulated during mESC differentiation. Oct4P4 lncRNA forms a complex with the SUV39H1 HMTase to direct the imposition of H3K9me3 and HP1α to the promoter of the ancestral Oct4 gene, located on chromosome 17, leading to gene silencing and reduced mESC self-renewal. Targeting Oct4P4 expression in primary mouse embryonic fibroblasts causes the re-acquisition of self-renewing features of mESC. We demonstrate that Oct4P4 lncRNA plays an important role in inducing and maintaining silencing of the ancestral Oct4 gene in differentiating mESCs. Our data introduces a sense pseudogene-lncRNA-based mechanism of epigenetic gene regulation that controls the cross-talk between pseudogenes and their ancestral genes.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis