MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Inhibition of Rho-associated kinase relieves C5a-induced proteinuria in murine nephrotic syndrome.
Inhibition of Rho-associated kinase relieves C5a-induced proteinuria in murine nephrotic syndrome.
Cellular and molecular life sciences : CMLS (2015-03-21)
I-Jung Tsai, Chia-Hung Chou, Yao-Hsu Yang, Wei-Chou Lin, Yen-Hung Lin, Lu-Ping Chow, Hsiao-Hui Lee, Pei-Gang Kao, Wan-Ting Liau, Tzuu-Shuh Jou, Yong-Kwei Tsau
ABSTRACT
Childhood nephrotic syndrome is mainly caused by minimal change disease which is named because only subtle ultrastructural alteration could be observed at electron microscopic level in the pathological kidney. Glomerular podocytes are presumed to be the target cells whose protein sieving capability is compromised by a yet unidentified permeability perturbing factor. In a cohort of children with non-hereditary idiopathic nephrotic syndrome, we found the complement fragment C5a was elevated in their sera during active disease. Administration of recombinant C5a induced profound proteinuria and minimal change nephrotic syndrome in mice. Purified glomerular endothelial cells, instead of podocytes, were demonstrated to be responsible for the proteinuric effect elicited by C5a. Further studies depicted a signaling pathway involving Rho/Rho-associated kinase/myosin activation leading to endothelial cell contraction and cell adhesion complex breakdown. Significantly, application of Rho-associated kinase inhibitor, Y27632, prevented the protein leaking effects observed in both C5a-treated purified endothelial cells and mice. Taken together, our study identifies a previously unknown mechanism underlying nephrotic syndrome and provides a new insight toward identifying Rho-associated kinase inhibition as an alternative therapeutic option for nephrotic syndrome.
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)