MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Ultra-high-performance liquid chromatography electrospray ionization tandem mass spectrometry for accurate analysis of glycerophospholipids and sphingolipids in drug resistance tumor cells.
Ultra-high-performance liquid chromatography electrospray ionization tandem mass spectrometry for accurate analysis of glycerophospholipids and sphingolipids in drug resistance tumor cells.
Journal of chromatography. A (2015-01-24)
Lin Li, Linlin Wang, Dihua Shangguan, Yanbo Wei, Juanjuan Han, Shaoxiang Xiong, Zhenwen Zhao
ABSTRACT
Glycerophospholipids and sphingolipids are important signaling molecules which are involved in many physiological and pathological processes. Here we reported an effective method for accurate analysis of these lipids by liquid chromatography electrospray ionization tandem mass spectrometry (LC-ESI-MS/MS). The methanol method was adopted for extraction of lipids due to its simplicity and high efficiency. It was found that two subclasses of sphingolipids, sulfatide (ST) and cerebroside (CB), were heat labile, so a decreased temperature in the ion source of MS might be necessary for these compounds analysis. In addition, it was found that the isobaric interferences were commonly existent, for example, the m/z of 16:0/18:1 PC containing two (13)C isotope being identical to that of 16:0/18:0 PC determined by a unit mass resolution mass spectrometer; therefore, a baseline separation of interferential species was required to maintain selectivity and accuracy of analysis. In this work, an ultra-high-performance liquid chromatography (UHPLC)-based method was developed for separation of interferential species. Moreover, in order to deal with the characteristics of different polarity and wide dynamic range of glycerophospholipids and sphingolipids in biological systems, three detecting conditions were combined together for comprehensive and rational analysis of glycerophospholipids and sphingolipids. The method was utilized to profile glycerophospholipids and sphingolipids in drug resistant tumor cells. Our results showed that many lipids were significantly changed in drug resistant tumor cells compared to paired drug sensitive tumor cells. This is a systematic report about the isobaric interferences and heat labile compounds interferences when analyzing glycerophospholipids and sphingolipids by ESI-MS/MS, which aids in ruling out one potential source of systematic error to ensure the accuracy of analysis.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
N-Palmitoyl-D-sphingosine, ≥98.0% (TLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer