Solvents
We are committed to providing you with the right solvent for your specific application. All our products undergo stringent controls and continuous development to meet your exacting requirements. Our comprehensive portfolio of solvents can be found under three of our Portfolio Brands: Supelco® products for analytical chemistry, Sigma-Aldrich® materials for lab and production, SAFC® products for biopharmaceutical and pharmaceutical development and manufacturing. All our products are designed to fit your application requirements and are available in a variety of sizes, packaging materials, and with the supporting documentation you need.
Related Resources
- Brochure: Safety Connects
We invest heavily in safe and innovative solvent handling technology and product development, focusing on customer requirements and offering a wide range of products and accessories. Our primary goal is to ensure user safety and the reliability of analytical results in your laboratory.
- Brochure: EMSURE® | EMPARTA® | EMPLURA® Inorganics & Solvents for Classical Analysis
We provide scientists with best-in-class portfolio particularly for lab applications. Our Life Science portfolio comprises more than 300,000 products, served to you in 66 countries around the world.
- Brochure: SupraSolv® Solvents for Gas Chromatography
As the world’s leading supplier of high-purity solvents, we offer a full range of products for every gas chromatography application in the laboratory - including highly sensitive pesticide and dioxin analyses. Our SupraSolv® and SupraSolv® hypergrade solvents are developed specifically for sensitive detection processes in residue and environmental analysis.
- Brochure: NMR Solvents
One of our core specialties is NMR, for which we produce premium quality solvents, standards, CRMs, and stable isotope reagents, as well as dedicated NMR tubes and accessories.
- Brochure: Sustainable Solvent Alternatives
The products we create help our customers improve people’s lives every day. But we realize that everything we make also has an environmental impact. That’s why we are committed to continually enhancing the sustainability of our products, and adopting environmentally friendly chemical processes. What’s more, we strive to make your dai
- Brochure: API Development and Manufacturing Services, Formulation Products, and Process Chemicals
Arcofolin™ offers supplementation comparable to enriching your diet with natural folate. As the monosodium salt of L-5-Methyltetrahydrofolic acid, it is the pure and crystalline form of the naturally occuring predominant form of folate.
- Brochure: Formulation Portfolio: Make Your Choice
In pharmaceutics, the regulatory world is everchanging. Keeping up with the latest requirements while not losing track of the market itself is definitely not an easy task to fulfill.
- Flyer: Advanced LC-MS Solvents
Our new range of advanced UHPLC-MS LiChrosolv® solvents have been developed to exceed all expectations, providing rapid and reliable results in both ESI/APCI positive and negative ionization modes.
- Brochure: The Future of Solvents - Bio Renewable
These solvents are free of the many by-products of petroleum manufacturing, such as benzene, aldehydes, and ethers. What’s more, each solvent is verified as renewable through ASTM testing (ASTM Standard D6866), which verifies the percentage of new carbon.
- Brochure: Reliable Research Must-Have Lab Reagents for Everyday Use
This comprehensive guide explains the differences between the grades and outlines common applications for each specification – to make your decisions easier, and your work smoother.
- Brochure: Sure/Seal™ System for Anhydrous Solvents
Choose from the largest range of high-quality anhydrous solvents with exceptionally low water levels. Rest assured that each product is perfectly protected with our innovative, moisture-inhibiting Sure/Seal™ system.
- Support: HPLC Method Transfer Calculator
Calculate the saving in run time and solvent consumption when transferring a method from HPLC conditions to UHPLC conditions. Obtain guidance on change in back pressure as well as how to adapt injection volume and gradient conditions.
Solvents for Instrumental analysis
We are committed to providing our customers with the widest selection of high purity solvents, specifically designed to deliver the ultimate performance for UHPLC-MS, LC-MS, HPLC, GC, and UV Instrumental Analysis. For this purpose, we offer two premium brands: Supelco® for regulated/accredited application fields and Sigma-Aldrich® for research application fields. LiChrosolv®, OmniSolv® (only in the US and Canada), SupraSolv®, and Uvasol® solvents are now residing under the Supelco® brand portfolio. Our large portfolios of analytical solvents allow reliable, accurate and reproducible results without the need for costly purification and repeated analysis.
LiChrosolv® solvents are ideal for analytical HPLC and LC-MS applications. These high-performance solvents offer excellent resolution, sensitivity, and optimal peak baseline separation. Their excellent UV transmittance, low particle counts, low acid and alkalinity, and low evaporation residue levels ensure reproducible separations. Microfiltered at 0.2 µm, they are available in both isocratic and gradient grades, with the latter minimizing gradient effects. The LiChrosolv® hypergrade solvents are suited for fast chromatography and LC-MS due to their low ionic background and minimal ion suppression. They meet modern LC-MS ionization standards while maintaining good repeatability and ionization efficiency. SupraSolv® solvents are ideal for all gas chromatography lab applications and prepared for use with various detector systems (ECD, FID & MS) or headspace sampling. Uvasol® solvents are specially designed for applications that demand solvents of the highest spectral purity and are widely used in IR, UV and fluorescence spectroscopy.
Solvents for classical analysis, production, and purification
We strive to tailor our comprehensive range of solvents to customer requirements. EMSURE® solvents for analysis are our premium grade suitable for a broad spectrum of classical lab applications including regulated and highly demanding applications. Most EMSURE® grade solvents meet or even exceed the requirements from ACS, but also from Reag. Ph. Eur. and ISO regulations. EMPARTA® is a high-quality solvent grade that fulfills the requirements of the ACS and can be used for a wide range of standard analytical applications. Further, EMPLURA® grade solvents are a cost-effective alternative for basic applications such as preparative lab work, production and cleaning.
Prepsolv® solvents are tailored to the requirements of preparative chromatography to facilitate scale-up from analytical to preparative separations. For RNA/DNA and peptide synthesis we offer a special range of solvents providing highest purity, as well as high and constant dryness. Furthermore, our solvents offer more flexibility, thanks to a wide range of packaging solutions including bottles and bulk containers up to 1000 liter.
Our premium-grade solutions, EMSURE® solvents, are designed for rigorous and regulated laboratory applications. They provide refined impurity profiles and enhanced purity, water content, and evaporation residue values. They meet or exceed the standards set by ACS, Reag. Ph. Eur., and ISO for pharmaceutical quality control.
Solvent for General Chemistry, Chemical Synthesis, Production and Biotech Application
Our ReagentPlus® solvents have a guaranteed purity of ≥98.5% for applications that require higher purities, such as specialty organic synthesis, preparative scale liquid chromatography or glass washing. ReagentPlus® solvents are available in a wide variety of containers. The ACS solvents can be used for ACS applications or for general procedures that require stringent quality specifications. Furthermore, Sigma-Aldrich® brand portfolio offers high-purity anhydrous solvents with extremely low water levels specifically manufactured for moisture sensitive organic and biotech applications.
NMR Solvents
We provide the widest range of NMR solvents with excellent chemical purity and the highest isotopic enrichment possible. To ensure the finest quality, we consistently review and improve our methods for solvent purification and water content reduction. All of our NMR solvents undergo strict quality control testing during production and packaging. For more specific requirements, we support you with the convenience of customized products and packaging. Our glass bottles, ampules, and septum bottles for NMR solvents are available in large range of configurations and sizes.
Biopharmaceutical and pharmaceutical formulation and production products
In pharmaceutics, the regulatory world is everchanging. Keeping up with the latest requirements while not losing track of the market itself is definitely not an easy task to fulfill. And as the pressure caused by generics and biosimilars making their way into the market keeps rising, it becomes more and more important to focus on what really matters: Quality and the right partner throughout your product journey.
We support you in every step of development, scale-up, and production with services that range from process optimization to regulatory affairs support. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation.