[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Solid Phase Extraction (SPE)](https://www.sigmaaldrich.com/US/en/applications/analytical-chemistry/sample-preparation/solid-phase-extraction)Pesticide Analysis in Turmeric by LC/MS/MS & GC/MS/MS
# Pesticide Analysis in Turmeric by LC/MS/MS & GC/MS/MS
__Katherine K. Stenerson, Principal R&D Scientist__
[Article from Analytix Reporter - Issue 1](https://www.sigmaaldrich.com/US/en/collections/analytix-reporter)
## Introduction
Turmeric is a plant indigenous to south Asia, with a majority of its production coming from India. The rhizome of the plant is used to produce powdered turmeric, which is used in foods, cosmetics, and some medicines. It is also an essential constituent of curry, which is a mixture of spices used extensively in Indian cooking. Turmeric has also been used in traditional medicines for thousands of years and recently has garnered attention for studies showing its potential antioxidant, anti-inflammatory, antimutagenic, antimicrobial, and anticancer properties.
Pesticide residue testing of turmeric and other spices is required by many countries. For example, Canada has set maximum residue limits for 42 different pesticides in turmeric root. The U.S. EPA has set tolerance limits for a variety of pesticides in root and tuberous vegetables, of which turmeric is included.
Turmeric contains more than 100 different components, with two of the main constituents being curcumin and volatile oils. Curcumin gives turmeric its distinctive yellow/orange color, while the volatile oils consist primarily of terpenes. Turmeric also contains some fats; specifically, sterols and fatty acids. This complex composition makes extracts produced from turmeric a challenge in the chromatographic analysis of pesticides, as residual pigments and oils can contaminate both GC/MS and LC/MS systems.
When dealing with very high background samples such as turmeric, standard QuEChERS cleanup may not offer enough capacity. For better cleanup, solid phase extraction (SPE), including dual-layer cartridges, can be used. These cartridges often contain graphitized carbon black (GCB) in the top bed and primary-secondary amine (PSA) in the bottom bed. PSA retains acidic interferences such as fatty acids. GCB removes planar molecules such as pigments and sterols. Common GCBs, however, will retain all molecules with planar structures, including some pesticide analytes such as hexachlorobenzene. To increase recoveries of these pesticides, toluene is normally added to the elution solvent. However, there are issues associated with the use of toluene. It can affect the ability of the PSA to retain fatty acids, and its presence in the final extract is problematic for HPLC analysis.
In this application, a different dual-layer SPE cartridge was used in the cleanup of extracts of turmeric powder prior to pesticide analysis by GC/MS/MS and LC/MS/MS. This cartridge, the Supelclean™ Ultra 2400, was designed for the cleanup of acetonitrile extracts made from difficult matrices such as dry commodities (spices, tea, etc.) prior to pesticide residue analysis. The top bed consists of a mixture of PSA, C18 and a graphitized, spherical carbon known as Graphsphere™ 2031. This carbon was engineered to remove sufficient pigmentation while allowing for better recoveries of planar compounds, without the need for toluene in the elution solvent. The bottom layer of the cartridge contains Z-Sep, a zirconia-coated silica. Z-Sep removes oily residues and provides additional retention of some pigments. The combination of these sorbents in an SPE format offers more capacity than QuEChERS cleanup, and compared to traditional GCB/PSA dual layer cartridges, does not require the use of toluene in the elution solvent to recover planar pesticides.
## Experimental
Turmeric powder was obtained from a local grocery store. Samples were spiked at 100 ng/g with the pesticides listed in __Tables 1 and 2__. Sample extracts were prepared and cleaned following the procedures in __Figure 1__. A set of 3 spiked samples and 1 unspiked (blank) were prepared and analyzed for each set of pesticides. Analysis was done by GC/MS/MS and LC/MS/MS using the conditions listed in __Tables 3 and 4__ (with MS/MS transitions shown in __Tables 1 and 2__). Quantitation was performed against multi-point calibration curves prepared in unspiked turmeric extract (after cleanup). Recoveries were calculated as the average of the three spiked replicates, less anything found in the unspiked extract. No internal standards were used, thus the values reported represent absolute recoveries.
## Results and Discussion
### Background
Prior to cleanup, the extract appeared orange-brown in color with a yellow oily residue (__Figure 2__). After cleanup for both LC and GC, the extracts appeared substantially lighter and clearer. __Figures 3 and 4__ show a comparison between extracts at the same level of dilution with and without cleanup. The LC extract (in 80% aqueous) was almost devoid of color, with very little cloudiness. The extract for GC analysis was a pale yellow color, with substantially less oily residue. Full scan GC/MS analyses of GC extracts are shown in __Figure 5__ as total ion chromatograms (TICs). The peak pattern is similar between the two, with the main peaks consisting primarily of terpenes. These compounds are easily volatilized in the GC inlet, and do not pose issues with system contamination, however they can interfere with mass spectral detection, requiring the use of MS/MS for selectivity. The overall amplitude of the peaks was less after cleanup, as is shown by a 21% reduction in the peak area sums for each in __Figure 5.__
| | | | | |
|----------------------|---------|----|---------|----|
| | MRM 1 | CE | MRM 2 | CE |
| Alachlor | 188/160 | 10 | 188/130 | 40 |
| Aldrin | 263/193 | 35 | 263/191 | 35 |
| γ-BHC | 183/147 | 15 | 181/145 | 5 |
| Azinphos-methyl | 160/77 | 15 | 132/77 | 15 |
| Chloropyrifos | 314/286 | 5 | 314/258 | 15 |
| Chloropyrifos-methyl | 286/93 | 20 | 288/93 | 20 |
| Cypermthrins | 165/91 | 10 | 163/91 | 10 |
| 4,4'-DDT | 235/199 | 15 | 235/165 | 25 |
| Diazinon | 199/135 | 15 | 137/84 | 10 |
| Dichlorvos | 185/93 | 25 | 145/109 | 25 |
| Dimethoate | 125/79 | 20 | 93/63 | 10 |
| Disulfoton | 88/60 | 5 | 88/59 | 15 |
| Endosulfan β | 241/206 | 15 | 241/170 | 30 |
| Endosulfan-α | 241/206 | 15 | 241/170 | 30 |
| Ethion | 231/129 | 20 | 121/65 | 10 |
| Fenitrothion | 277/125 | 20 | 277/109 | 20 |
| Heptachlor | 274/239 | 15 | 272/237 | 15 |
| Hexachlorobenzene | 284/249 | 20 | 284/214 | 35 |
| Iprodione I | 314/56 | 35 | 187/124 | 25 |
| Iprodione II | 316/56 | 35 | 187/124 | 25 |
| Malathion | 173/99 | 15 | 158/125 | 5 |
| Metalaxyl | 234/174 | 10 | 234/146 | 20 |
| Methoxychlor | 227/169 | 30 | 227/141 | 30 |
| Mevinphos | 192/127 | 25 | 192/109 | 25 |
| Parathion-methyl | 233/109 | 10 | 124/47 | 10 |
| Permethrins | 183/168 | 10 | 183/165 | 10 |
| Phenthoate | 274/125 | 15 | 274/121 | 10 |
| Phorate | 260/75 | 5 | 231/129 | 25 |
| Phosalone | 182/102 | 15 | 182/75 | 30 |
| Pirimiphos-methyl | 290/151 | 20 | 290/125 | 25 |
| Profenophos | 339/269 | 15 | 339/188 | 15 |
| Quintozene | 295/237 | 20 | 237/143 | 30 |
| Vinclozolin | 212/145 | 30 | 187/124 | 20 |
Table 1 Pesticides Studied in Turmeric Powder by GC/MS/MS Analysis
| | | | | |
|-------------------------|--------------|----------|--------|--------------|
| | MRM | Frag (V) | CE (V) | Cell Acc (V) |
| Acephate | 184/143 | 70 | 0 | 5 |
| Acetamiprid | 223.1/126 | 80 | 27 | 2 |
| Boscalid (Nicobifen) | 343/307.1 | 145 | 16 | 6 |
| Carbendazim (Azole) | 192.1/160.1 | 105 | 16 | 2 |
| Chlorbufam | 224/172.02 | 120 | 5 | 3 |
| Cycluron | 199.2/72 | 120 | 20 | 2 |
| Diflubenzuron | 311/158 | 80 | 8 | 2 |
| Fenoxanil | 329.08/189 | 80 | 30 | 3 |
| Fosthiazate | 284/61 | 90 | 60 | 2 |
| Methabenzthiazuron | 222.1/165.1 | 90 | 12 | 2 |
| Methamidophos | 142/125 | 85 | 10 | 2 |
| Methomyl | 163.1/106 | 50 | 4 | 2 |
| Monocrotophos (Azodrin) | 224.1/193 | 65 | 0 | 5 |
| Nitralin | 346.11/304 | 100 | 10 | 3 |
| Oxamyl | 237.1/72 | 60 | 12 | 2 |
| Pirimicarb | 239.15/72.1 | 100 | 20 | 2 |
| Procymidon | 301/284* | 70 | 8 | 2 |
| Propaquizafop | 444.12/100.1 | 125 | 16 | 2 |
| Tetraconazole | 372/159 | 130 | 36 | 2 |
| Uniconazole-P | 292.1/125 | 135 | 40 | 2 |
Table 2Pesticides Studied in Turmeric Powder by LC/MS/MS Analysis * Ammonium adduct
| | |
|----------------|------------------------------------------------------------------------------------------|
| column: | SLB®-5ms, 30 m × 0.25 mm ID, 0.25 μm (28471-U) |
| oven: | 50 °C (2 min), 8 °C/min to 320 °C (5 min) |
| inj. temp.: | 250 °C |
| carrier gas: | helium, 1.4 mL/min, constant |
| detector: | MRM (table 3) |
| MSD interface: | 320 °C |
| injection : | 1μL, splitless (splitter open at 0.75 min) |
| liner: | 4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design (2879901-U) |
Table 3GC/MS/MS Run Conditions for the Analysis of Pesticides in Turmeric
column:Ascentis® Express C18, 10 cm × 2.1 mm ID, 2 μm
mobile phase:\[A] 5 mM ammonium formate, 0.1% formic acid in water;
\[B] 5 mM ammonium formate, 0.1% formic acid in methanol
gradient:95% A, 5% B held for 1 min; to 50% A in 3 min; to 100%
B in 8 min; held for 1 min;, to 95% A in 1.5 min; held at 95%
A for 1.5 min
flow rate:0.4 mL/min
detector:MRM (table 2)
injection:5 μL
Table 4 LC/MS/MS Run Conditions for the Analysis of Pesticides in Turmeric
__Figure 1.__Extraction and Cleanup Procedure Used for Turmeric Powder, GC and LC
[](#)
__Figure 2.__ Undiluted Acetonitrile Extract of Turmeric Powder Before Cleanup
[](#)
__Figure 3.__ Turmeric Extracts at the Same Dilution (167X total); Without Cleanup, and After Cleanup for LC/MS/MS Analysis
[](#)
__Figure 4.__ Turmeric Extracts at the Same Dilution (5X); Without Cleanup, and After Cleanup for GC/MS/MS Analysis
[](#)
## Pesticide Recovery and Reproducibility
The average recoveries obtained from spiked turmeric samples (n=3) are presented in __Table 5__. Of the 51 pesticides spiked, all except hexachlorobenzene had recovery of greater than 70%. Hexachlorobenzene, a pesticide with a planar structure, was recovered at 67% after cleanup. It should be noted that this was without using toluene in the elution solvent, as is necessary to obtain good recoveries from dual-layer cartridges containing graphitized carbon black. Although not shown here, higher recovery of hexachlorobenzene has been obtained by loading more turmeric extract (300 μL) on the Supelclean™ Ultra 2400 cartridge. This indicates that the presence of more matrix displaced the hexachlorobenzene, thus reducing its retention on the carbon. However the higher sample loading produced an extract with more color, a sign that the cleanup capacity of the cartridge had been reached or exceeded for this matrix.
| | | | |
|-------------------------|---------------|-----|----------|
| Pesticide | Avg. Recovery | RSD | Analysis |
| Alachlor | 99% | 23% | GC/MS/MS |
| Aldrin | 85% | 10% | GC/MS/MS |
| Azinphos-methyl | 89% | 11% | GC/MS/MS |
| γ-BHC | 83% | 8% | GC/MS/MS |
| Chloropyrifos | 96% | 12% | GC/MS/MS |
| Chloropyrifos-Methyl | 113% | 6% | GC/MS/MS |
| Cypermthrin (isomer 1) | 99% | 15% | GC/MS/MS |
| 4,4'-DDT | 95% | 8% | GC/MS/MS |
| Diazinon | 92% | 14% | GC/MS/MS |
| Dichlorvos | 78% | 31% | GC/MS/MS |
| DIsulfoton | 86% | 7% | GC/MS/MS |
| Endosulfan β | 86% | 35% | GC/MS/MS |
| Endosulfan-α | 92% | 23% | GC/MS/MS |
| Ethion | 97% | 7% | GC/MS/MS |
| Fenitrothion | 63% | 5% | GC/MS/MS |
| Heptachlor | 81% | 7% | GC/MS/MS |
| Hexachlorobenzene | 67% | 9% | GC/MS/MS |
| Iprodione (isomer 1) | 103% | 5% | GC/MS/MS |
| Malathion | 90% | 10% | GC/MS/MS |
| Metalaxyl | 86% | 21% | GC/MS/MS |
| Methoxychlor | 78% | 12% | GC/MS/MS |
| Mevinphos | 73% | 7% | GC/MS/MS |
| Parathion-Methyl | 88% | 8% | GC/MS/MS |
| Permethrin (isomer 1) | 104% | 24% | GC/MS/MS |
| Phenthoate | 89% | 7% | GC/MS/MS |
| Phorate | 82% | 10% | GC/MS/MS |
| Phosalone | 90% | 7% | GC/MS/MS |
| Pirimiphos-methyl | 74% | 3% | GC/MS/MS |
| Profenophos | 88% | 7% | GC/MS/MS |
| Quintozene | 75% | 8% | GC/MS/MS |
| Vinclozolin | 90% | 6% | GC/MS/MS |
| Acephate | 89% | 6% | LC/MS/MS |
| Acetamiprid | 102% | 4% | LC/MS/MS |
| Boscalid (Nicobifen) | 86% | 7% | LC/MS/MS |
| Carbendazim (Azole) | 106% | 7% | LC/MS/MS |
| Chlorbufam | 92% | 18% | LC/MS/MS |
| Cycluron | 103% | 5% | LC/MS/MS |
| Diflubenzuron | 101% | 5% | LC/MS/MS |
| Fenoxanil | 91% | 10% | LC/MS/MS |
| Fosthiazate | 95% | 4% | LC/MS/MS |
| Methabenzthiazuron | 96% | 4% | LC/MS/MS |
| Methamidophos | 85% | 5% | LC/MS/MS |
| Methomyl | 106% | 6% | LC/MS/MS |
| Monocrotophos (Azodrin) | 97% | 3% | LC/MS/MS |
| Nitralin | 124% | 55% | LC/MS/MS |
| Oxamyl | 104% | 3% | LC/MS/MS |
| Pirimicarb | 97% | 3% | LC/MS/MS |
| Procymidon | 91% | 13% | LC/MS/MS |
| Propaquizafop | 97% | 1% | LC/MS/MS |
| Tetraconazole | 98% | 2% | LC/MS/MS |
| Uniconazole-P | 103% | 19% | LC/MS/MS |
Table 5 Pesticide Recoveries and % RSD Values (n=3) for Spiked Replicates; Turmeric Spiked at 100 ng/g
__Figure 5.__GC/MS Scan Analyses of Turmeric Extracts Before and After Cleanup with Supelclean™ Ultra 2400 Shown with same Y-scale. Sum of area counts for all peaks is indicated with each.
## Recoveries Within Indicated Percent Relative
__Figure 6.__ Number of Pesticides with Average Standard Deviation (%RSD) Ranges After Cleanup with Supelclean™ Ultra 2400 (Recoveries from turmeric powder spiked at 100 ng/g.)
## Conclusion
A new cleanup method has been developed using the Supelclean™ Ultra 2400 dual-layer SPE cartridge. The selection of sorbents in this cartridge allows for cleanup of acetonitrile extracts of very difficult samples such as spices and other dry commodities. The Graphsphere™ 2031 carbon used in the upper layer removes/reduces pigmentation while still allowing for recovery of planar pesticides without the use of toluene in the elution solvent. Z-Sep sorbent in the bottom layer of the cartridge removes oils and some pigments, as was indicated in the cleanup of turmeric extracts for both GC and HPLC analysis. Suitable recoveries for a wide range of pesticides of different polarities and classes were obtained from turmeric extract, and minimal background interference was noted. In this work, a 1 mL Supelclean™ Ultra 2400 cartridge was used. A larger 3 mL version of the cartridge is also available which can accommodate a higher sample loading.
## Acknowledgements
The author would like to thank Richard Schriner and Bruce Morris of R.J. Hill Laboratories for their helpful discussions and input.
* * *
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
* * *
### References
1\.
Prasad S, Aggarwal B. 2011. Turmeric, the Golden Spice.263-288. [https://doi.org/10.1201/b10787-14](https://doi.org/10.1201/b10787-14)
2\.
2016\. Health Canada MRL database. \[Internet]. Available from: [http://www.hc-sc.gc.ca](http://www.hc-sc.gc.ca)
3\.
2016\. Electronic Code of Federal Regulations (eCFR) Title 40, Chapter 1, Subchapter E, Part 180. \[Internet]. Available from: [https://www.ecfr.gov/cgi-bin/text-idx?tpl=/ecfrbrowse/Title40/40cfr180\_main\_02.tpl](https://www.ecfr.gov/cgi-bin/text-idx?tpl=%2Fecfrbrowse%2FTitle40%2F40cfr180_main_02.tpl)
4\.
2011\. Chemical composition and product quality control of turmeric (Curcuma longa L.). \[Internet]. Available from: [https://core.ac.uk/download/pdf/72733173.pdf](https://core.ac.uk/download/pdf/72733173.pdf)
5\.
Shimelis O, Yang Y, Stenerson K, Kaneko T, Ye M. 2007. Evaluation of a solid-phase extraction dual-layer carbon/primary secondary amine for clean-up of fatty acid matrix components from food extracts in multiresidue pesticide analysis. Journal of Chromatography A. 1165(1-2):18-25. [https://doi.org/10.1016/j.chroma.2007.07.037](https://doi.org/10.1016/j.chroma.2007.07.037)
__Related Articles__
- [Determination of Formaldehyde and Acetaldehyde in Air Using DNPH Cartridges and Automated On-Line Desorption Followed by HPLC](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/environmental-testing-and-industrial-hygiene/air-testing/aldehydes-air-dnph)
- [Effect of Container Materials on Milk Off-Flavors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/gas-chromatography/container-materials)
- [Pesticide Residue Analysis in Ginger Powder with QuEChERS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/quechers/analysis-of-pesticide-residues-in-ginger-powder)
- [Analysis of Pesticide Residues in Pistachios Using QuEChERS Extraction and Cleanup with Supel™ QuE Z-Sep+](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/quechers/analysis-of-pesticide-residues-in-pistachios)
- [HybridSPE-Phospholipid Technology](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/hybridspe-precipitation-technology)
- [Impact of Mobile Phase Additives on LC-MS Sensitivity, Demonstrated using Spice Cannabinoids](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/lcms-spice-cannabinoids)
- [Solvent Analysis for 40 Per and Polyfluoroalkyl Substances](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/solvent-analysis-for-40-per-and-polyfluoroalkyl-substances)
- [Analysis of Drugs of Abuse in Urine](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/solid-phase-extraction/analysis-of-drugs-of-abuse-in-urine-after-cleanup-with-hlb)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Solid Phase Extraction (SPE) and QuEChERS](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-sample-preparation/spe-and-quechers)
- [Chromatography & Spectroscopy Reagents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-reagents/chromatography-spectroscopy-reagents)
- [HPLC Columns](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
- [Environmental Accounting Supplier Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/environmental-accounting-supplier-toolkit)
- [Renewable Electricity Toolkit](https://www.sigmaaldrich.com/US/en/life-science/ssbi/sustainability-toolkits/renewable-electricity-toolkit)
- [Crown Ethers and Cryptands for Advanced Applications](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/crown-ethers-cryptands-advanced-applications)
- [Purification of Extracellular Vesicles with Eshmuno® Q Resin and Natrix® Q Membrane](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/membrane-chromatography/purification-extracellular-vesicles)
- [Sterilizing filters for final sterile filtration](https://www.sigmaaldrich.com/US/en/technical-documents/product-supporting/pharma-and-biopharma-manufacturing/final-sterile-filtration-and-filling/sterilizing-grade-membrane-filters)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)