Organic Building Blocks
Although new catalysts and reagents are steadily discovered in synthetic chemistry, scientists continue to search for the most appropriate substrate for their research. It might be easy to overlook organic building blocks, but access to the right substrate is every bit as important as having the tools to transform it into the desired target.
Organic building blocks are organic functionalized molecules: the basic components for organic synthesis. They are used for bottom-up modular assembly of molecular architectures, such as supra-molecular complexes, metal-organic frameworks, organic molecular constructs, and nanoparticles. Thereby, they play a fundamental role in medicinal chemistry, organic chemistry, and material chemistry.
Use our structure search to find building blocks with specific chemical structures.
We offer a diverse array of organic building blocks:
|
|
Spark your chemical synthesis with our one-stop source of chemical reagents. Alkynes are critical substrates in Sonogashira coupling and click chemistry. Pyridines are extremely common building blocks for a wide range of synthetic targets. Aldehydes are perennially attractive building blocks due to their ability to easily react with a variety of nucleophiles. Hindered amines are classically very challenging to access with current synthetic methods with broad application in synthesis, drug discovery, and agriscience. By combining our core offering with a diverse suite of partners’ compounds, we have the industry’s most comprehensive selection of organic building blocks.
Related Resources
- Baran Diversinates™
Rapidly diversify (hetero)aromatic scaffolds for chemical industry needs amid resource and time constraints, ensuring efficiency.
- SuFEx: Sulfonyl Fluorides that Participate in the Next Click Reaction
In collaboration with K. Barry Sharpless and coworkers, we offer a variety of sulfonyl fluoride reagents that undergo a “Click II” reaction.
- Hydrosilylation Catalyst
Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.
- Asymmetric Epoxidation Using Shi Catalyst
Catalytic asymmetric epoxidation of alkenes has been the focus of many research efforts over the past two decades, the best known methods probably being those developed by Sharpless and Jacobsen-Katsuki.