Merck
  • Arsenic trioxide induces programmed cell death through stimulation of ER stress and inhibition of the ubiquitin-proteasome system in human sarcoma cells.
Arsenic trioxide induces programmed cell death through stimulation of ER stress and inhibition of the ubiquitin-proteasome system in human sarcoma cells.
Cancer letters (2014-12-03)
Hui-Wen Chiu, Yin-Chiu Tseng, Yung-Ho Hsu, Yuh-Feng Lin, Ning-Ping Foo, How-Ran Guo, Ying-Jan Wang
ABSTRAKT
Sarcoma is a rare form of cancer that differs from the much more common carcinomas because it occurs in a distinct type of tissue. Many patients of sarcoma have poor response to chemotherapy and an increased risk for local recurrence. Arsenic trioxide (ATO) is used to treat certain types of leukemia. Recently, data have revealed that ATO induces sarcoma cell death in several types of solid tumor cell lines. In the present study, we investigated whether ATO induces cancer cell death and elucidated the underlying anti-cancer mechanisms. Our results showed that ATO caused concentration- and time-dependent cell death in human osteosarcoma and fibrosarcoma cells. The types of cell death that were induced by ATO were primarily autophagy and apoptosis. Furthermore, ATO activated p38, JNK and AMPK and inhibited the Akt/mTOR signaling pathways. Specifically, we found that ATO induced endoplasmic reticulum (ER) stress and suppressed proteasome activation in two types of sarcoma cell lines. However, the level of proteasome inhibition in osteosarcoma cells was lower than in fibrosarcoma cells. Thus, we used combined treatment with ATO and a proteasome inhibitor to examine the antitumor activity in fibrosarcoma cells. The data indicated showed that the combination treatment of ATO and MG132 (a proteasome inhibitor) resulted in synergistic cytotoxicity. In a fibrosarcoma xenograft mouse model, the combined treatment significantly reduced tumor progression. Immunohistochemical studies revealed that combined treatment induced autophagy and apoptosis. In summary, our results suggest a potential clinical application of ATO in sarcoma therapy and that combined treatment with a proteasome inhibitor can increase the therapeutic efficacy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, tablet
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 5 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Informacje o cenach i dostępności nie są obecnie dostępne.
SAFC
Sodium chloride solution, 5 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Arsenic(III) oxide, ReagentPlus®, ≥99.0%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.