Merck
The presence of an RNA:DNA hybrid that is prone to slippage promotes termination by T7 RNA polymerase.
Journal of molecular biology (2014-07-01)
Vadim Molodtsov, Michael Anikin, William T McAllister
ABSTRAKT
Intrinsic termination signals for multisubunit bacterial RNA polymerases (RNAPs) encode a GC-rich stem-loop structure followed by a polyuridine [poly(U)] tract, and it has been proposed that steric clash of the stem-loop with the exit pore of the RNAP imposes a shearing force on the RNA in the downstream RNA:DNA hybrid, resulting in misalignment of the active site. The structurally unrelated T7 RNAP terminates at a similar type of signal (TΦ), suggesting a common mechanism for termination. In the absence of a hairpin (passive conditions), T7 RNAP slips efficiently in both homopolymeric A and U tracts, and we have found that replacement of the U tract in TΦ with a slippage-prone A tract still allows efficient termination. Under passive conditions, incorporation of a single G residue following a poly(U) tract (which is the situation during termination at TΦ) results in a "locked" complex that is unable to extend the transcript. Our results support a model in which transmission of the shearing force generated by steric clash of the hairpin with the exit pore is promoted by the presence of a slippery tracts downstream, resulting in alterations in the active site and the formation of a locked complex that represents an early step in the termination pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Supelco
Urea, analytical standard
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture