Merck
Antitumor activity of acriflavine in lung adenocarcinoma cell line A549.
Anticancer research (2014-11-05)
Chia-Jen Lee, Chia-Herng Yue, Yu-Jie Lin, Yu-Yu Lin, Shao-Hsuan Kao, Jer-Yuh Liu, Yieng-How Chen
ABSTRAKT
Aim/Materials and Methods: In order to develop better drugs against non-small cell lung cancer (NSCLC), we screened a variety of compounds and treated the human lung adenocarcinoma cell line A549 with different drug concentrations. We then examined the cell viability using the MTT assay. Data show that a new candidate drug, acriflavine (ACF), suppresses the viability of A549 cells in a concentration- and time-dependent manner. Flow cytometry analysis revealed that ACF significantly caused cell growth arrest in the G2/M phase on A549 cells. Moreover, ACF decreased Bcl-2 expression and increased Bax expression. The content of cleaved poly(ADP-ribose)polymerase-1 (PARP-1) and caspase-3 are significantly increased. These findings suggest that ACF is cytotoxic against A549 cells and suppresses A549 cells growth through the caspase-3 activation pathway. In the in vivo test, nude mice bearing A549 cells xenografts by intravenous injection were randomly assigned into two groups: control and experimental group. Treatment was initiated 10 days after implantation and intraperitoneal injection of 0.9% normal saline or 2 mg/kg of ACF was continued daily for five weeks. ACF treatment significantly decreased tumor size and tumor spots on lung surface of tumor-bearing mice. ACF can inhibit cell growth in A549 cells. Our results may assist on the delineation of the mechanism(s) leading to NSCLC cell growth inhibition and provide a new antitumor strategy against NSCLC.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine, ≥96.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Acridine, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
1-Aminoanthracene, technical grade, 90%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acridine Yellow G, Dye content 90 %
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)