Merck
The role of nitric oxide synthase in an early phase Cd-induced acute cytotoxicity in MCF-7 cells.
Biological trace element research (2014-12-17)
Lingying Zhong, Lumei Wang, Lurong Xu, Qunlu Liu, Linlei Jiang, Yuee Zhi, Wei Lu, Pei Zhou
ABSTRAKT
Literature to date has confirmed that cadmium (Cd) can accomplish its toxic effects via the free radical-induced damage, but Cd itself cannot generate free radicals directly. Nitric oxide (NO) is a fundamental molecule that interplays with reactive oxygen species (ROS), which may be associated with the Cd-induced cytotoxicity. However, the role of nitric oxide synthase (NOS) in an early phase Cd-induced acute cytotoxicity and its interaction has not been studied. In this report, we provide data showing that CdCl2 (10 μM, 100 μM, 1 mM) could modulate NOS activity in terms of NO production which was first suppressed with the release of Ca(2+) and Zn(2+), then induced with the transcriptional and translational activation of the three NOS isoforms in a possible feedback manner. The ROS level in cells was increased after CdCl2 exposure. By using the free radical scavenger N-acetyl-L-cysteine (LNAC) or the NOS activity inhibitor N(G)-methyl-L-arginine (LNMMA), it was demonstrated that NOS played a critical role on the Cd-induced ROS generation. The Cd-induced cytotoxicity was associated with the NOS-mediated oxidative stress in MCF-7 cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylocysteina, United States Pharmacopeia (USP) Reference Standard