Merck
  • A systematic comparison of the impact of inflammatory signaling on absorption, distribution, metabolism, and excretion gene expression and activity in primary human hepatocytes and HepaRG cells.
A systematic comparison of the impact of inflammatory signaling on absorption, distribution, metabolism, and excretion gene expression and activity in primary human hepatocytes and HepaRG cells.
Drug metabolism and disposition: the biological fate of chemicals (2014-12-07)
Marcus Klein, Maria Thomas, Ute Hofmann, Daniel Seehofer, Georg Damm, Ulrich M Zanger
ABSTRAKT
Inflammatory processes are associated with compromised metabolism and elimination of drugs in the liver, largely mediated by proinflammatory cytokines, such as interleukin-6. The Hepa-RG cell line is an established surrogate for primary human hepatocytes (PHH) in drug metabolism and toxicity studies. However, the impact of inflammatory signaling on HepaRG cells has not been well characterized. In this study, the response of primary human hepatocytes and HepaRG cells to interleukin (IL)-6 was comparatively analyzed. For this purpose, broad-spectrum gene expression profiling, including acute-phase response genes and a large panel of drug-metabolizing enzyme and transporter (DMET) genes as well as their modifiers and regulators, was conducted in combination with cytochrome P450 (P450) activity measurements. Exposure of PHH and HepaRG cells to IL-6 resulted in highly similar coordinated reduction of DMET mRNA, including major ATP-binding cassette transporters (ABCs), P450s, glutathione S-transferases (GSTs), uridine diphosphate glucuronosyltransferases (UGTs), and solute carriers (SLCs). Enzyme activities of CYP1A2, CYP2B6, CYP2C8, CYP2C9, CYP2C19, and CYP3A4 were reduced upon 48-72 hours exposure to IL-6 in PHH and HepaRG. However, although these effects were not significant in PHH due to large interindividual donor variability, the impact on HepaRG was more pronounced and highly significant, thus emphasizing the advantage of HepaRG as a more reproducible model system. Exposure of HepaRG cells to interleukin-1β and tumor necrosis factor α resulted in similar effects on gene expression and enzyme activities. The present study emphasizes the role of proinflammatory cytokines in the regulation of drug metabolism and supports the use of HepaRG in lieu of PHH to minimize subject variability.
MATERIAŁY
Numer produktu
Marka
Opis produktu
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SAFC
L-Glutamine
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications