Merck
Rapamycin inhibits mSin1 phosphorylation independently of mTORC1 and mTORC2.
Oncotarget (2015-03-05)
Yan Luo, Lei Liu, Yang Wu, Karnika Singh, Bing Su, Nan Zhang, Xiaowei Liu, Yangmei Shen, Shile Huang
ABSTRAKT
Current knowledge indicates that the mammalian target of rapamycin (mTOR) functions as two complexes, mTORC1 and mTORC2, regulating cell growth, proliferation, survival, differentiation, and motility. Recently mSin1 has been identified as a critical component of mTORC2, which is essential for phosphorylation of Akt and other signaling molecules. Studies have shown that rapamycin inhibits phosphorylation of mSin1. However, the underlying mechanism is unknown. Here we found that rapamycin inhibited phosphorylation of mSin1 potently and rapidly. Expression of rapamycin-resistant mutant of mTOR (mTOR-T), but not rapamycin-resistant and kinase dead mutant of mTOR (mTOR-TE), prevented rapamycin from inhibiting mSin1 phosphorylation, suggesting that rapamycin-induced dephosphorylation of mSin1 is mTOR-dependent. Surprisingly, ectopic expression of rapamycin-resistant and constitutively active p70 S6 kinase 1 (S6K1) did not confer resistance to rapamycin-induced dephosphorylation of mSin1. Furthermore, disruption of mTORC1 and mTORC2 by silencing raptor and rictor, respectively, or downregulation of S6K1 or Akt did not induce the dephosphorylation of mSin1 as rapamycin did. However, silencing mTOR or mLST8 mimicked the effect of rapamycin, inhibiting mSin1 phosphorylation. Our findings suggest that rapamycin inhibits mSin1 phosphorylation, which is independent of mTORC1 and mTORC2, but is possibly dependent on a new mTOR complex, which at least contains mTOR and mLST8.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%