Merck
Uptake and cytotoxicity of docetaxel-loaded hyaluronic acid-grafted oily core nanocapsules in MDA-MB 231 cancer cells.
Pharmaceutical research (2014-03-20)
Ibrahima Youm, Vivek Agrahari, James B Murowchick, Bi-Botti C Youan
ABSTRAKT
It is hypothesized that docetaxel (Doc)-loaded hyaluronic acid (HA)-polyethylene glycol/poly(ε-caprolactone)-grafted oily core nanocapsules (NCs) can enhance the drug cytotoxicity and uptake in CD44 expressing breast cancer (BC) cells (MDA-MB 231). NCs were prepared, optimized and characterized by dynamic light scattering, transmission electron microscopy (TEM), and powder X-ray diffraction (PXRD). In vitro cytotoxicity tests [MTS, level of reactive oxygen species (ROS) and level of reduced glutathione (GSH)] were performed in BC cells. The contribution of CD44 to the NCs cellular uptake was elucidated using an anti CD44 antibody blockage and a CD44 negative NIH3T3 cell line. The optimum formulation of Doc-loaded HA oily core NCs had respective mean diameter, polydispersity, and drug encapsulation efficiency of 224.18 nm, 0.32, and 60.38%. The NCs appeared spherical with low drug crystallinity, while the drug release data fitted to first order equation. Compared to that of ungrafted NCs, the cytotoxicity of Doc-loaded HA-grafted NCs was significantly enhanced (p<0.05). A decrease of the intracellular level of ROS was reversely correlated with that of GSH. Interestingly, the cellular internalization of HA-grafted NCs mediated CD44 was dramatically enhanced (3 to 4-fold) with respect to the absence of specific biomarker or targeting ligand. The use of HA-grafted NCs enhanced the selective drug payload, cytotoxicity and uptake in MDA-MB 231 cells. Therefore, it could be a promising template for safe and effective delivery of Doc and similar chemotherapeutic agents in cancer cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Chlorobimane
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
USP
Rozpuszczalnik resztkowy klasy 2 - acetonitryl, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%